Search
Search
| Gene Symbol: Tcp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290712.1 | NP_001277641.1 | 2 - 3 | 432 | 89 | |
| NM_013686.4 | NP_038714.2 | 3 - 4 | 518 | 89 | ||
| GenBank mRNA | AK149611.1 | - | 3 - 4 | 383 | 89 | |
| AK149755.1 | - | 3 - 4 | 384 | 89 | ||
| AK151068.1 | - | 3 - 4 | 383 | 89 | ||
| AK151445.1 | - | 2 - 3 | 322 | 89 | ||
| AK152641.1 | - | 3 - 4 | 375 | 89 | ||
| AK152759.1 | - | 3 - 4 | 384 | 89 | ||
| AK153037.1 | - | 3 - 4 | 384 | 89 | ||
| AK153430.1 | - | 2 - 3 | 322 | 89 | ||
| AK165665.1 | - | 3 - 4 | 385 | 89 | ||
| AK166828.1 | - | 3 - 4 | 382 | 89 | ||
| AK166966.1 | - | 3 - 4 | 406 | 89 | ||
| AK167529.1 | - | 3 - 4 | 383 | 89 | ||
| BC003809.1 | - | 3 - 4 | 366 | 3599670 | 89 | |
| CN725473.1 | - | 2 - 3 | 432 | 89 | ||
| D90344.1 | - | 3 - 4 | 333 | 89 | ||
| M12899.1 | - | 3 - 4 | 341 | 89 | ||
Set Membership: |
|