Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00496787_s1
          See other KRTAP15 GE Assays ›
          Gene Symbol:
          Krtap15
          Gene Name:
          keratin associated protein 15
          Gene Aliases:
          Krtap15-1, Pmg-2, Pmg2
          Chromosome Location:
          Chr.16: 88829009 - 88829844 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00496787_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Krtap15
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_013713.1 NP_038741.1 1 617 64
          GenBank mRNA AK132534.1 - 1 633 64

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1600000966.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Krtap15

          Panther Classification:

          Molecular Function -

          structural protein

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P498429 NM_013713.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P498429 AK132534.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P498429 BC116219.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P498429 NM_013713.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P498429 AK132534.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P498429 BC116219.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 679629 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P110899 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P498429 NM_013713.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P498429 AK132534.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P498429 BC116219.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P498429 NM_013713.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P498429 AK132534.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P498429 BC116219.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30948195 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30948189 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30948182 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30948183 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30948178 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30948194 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30948199 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30948196 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30948179 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30948192 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30948185 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30948200 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30948198 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P110899 - -
          Whole Mouse Genome G4122A A_51_P498429 NM_013713.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P498429 AK132534.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P498429 BC116219.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P498429 NM_013713.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P498429 AK132534.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P498429 BC116219.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P109092 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P110899 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30948130 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30948162 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30948150 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30948142 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30948153 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30948172 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30948144 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30948177 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30948167 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30948149 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30948212 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30948110 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30948157 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30948135 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30948199 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30948115 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30948189 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30948207 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30948124 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30948219 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30948198 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30948195 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30948155 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30948116 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30948161 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30948126 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30948128 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30948159 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30948118 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30948170 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30948140 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30948107 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30948139 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30948230 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30948141 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30948165 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30948218 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30948134 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30948166 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30948174 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30948192 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30948122 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30948217 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30948145 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30948208 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30948222 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30948183 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30948203 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30948228 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30948200 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30948178 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30948209 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30948196 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30948143 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30948147 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30948179 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30948175 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30948215 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30948204 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30948185 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30948205 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30948158 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30948168 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30948194 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12833085 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30948174 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30948215 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30948202 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30948212 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30948200 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30948209 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30948217 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30948196 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12485158 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30948211 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12638573 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30948172 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30948195 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30948185 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30948205 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30948168 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30948167 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12813558 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30948183 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30948189 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30948177 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12598121 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30948192 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30948170 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30948166 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30948219 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12813544 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30948220 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30948175 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30948208 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30948198 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30948207 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12833086 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30948178 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12833079 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10436755 - -
          Affymetrix Mouse Gene 1.0 ST Array 10436756 - -
          Affymetrix Mouse Gene 1.0 ST Array 10436753 - -
          Affymetrix Mouse Gene 1.0 ST Array 10436754 - -
          Affymetrix Mouse Gene 1.0 ST Array 10436757 - -
          Affymetrix HT MG-430 PM Array Plate 1419507_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10436752 - -
          Mouse Genome 430 2.0 Array 1419507_PM_at - -
          Mouse Genome 430 2.0 Array 1419507_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 26560_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 26560_at - -
          Affymetrix Mouse Clariom S Array PSR1600007583.mm.1 NM_013713.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600007583.mm.1 NM_013713.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600007583.mm.1 TC1600000966.mm.1 MTA TC
          Mouse Expression Set 430 1419507_at - -
          Mouse Expression Set 430 1419507_PM_at - -
          Mouse Genome 430A 2.0 Array 1419507_PM_at - -
          Mouse Genome 430A 2.0 Array 1419507_at - -
          Affymetrix HT Mouse Genome 430A Array 1419507_at - -
          Affymetrix HT Mouse Genome 430A Array 1419507_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 436347 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2470722 - -
          Illumina mouse-6 v1.0 expression beadchip 2470722 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002470722 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1236982 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1236982 - -
          mouse-6 v1.1 (Illumina) scl1157.1.1_232-S - -
          Illumina Sentrix mouse-6.v1 bead array 2470722 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl1157.1.1_232-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2470722 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1236982 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1236982 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline