Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00497168_g1
          See other BLOC1S1 GE Assays ›
          Gene Symbol:
          Bloc1s1
          Gene Name:
          biogenesis of lysosomal organelles complex-1, subunit 1
          Gene Aliases:
          AI839753, BLOS1, Gcn5l1
          Chromosome Location:
          Chr.10: 128919220 - 128923547 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00497168_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Bloc1s1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_015740.3 NP_056555.2 1 - 2 145 116
          GenBank mRNA AK003035.1 - 1 - 2 146 116
          BC034662.1 - 1 - 2 89 5359972 116

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1000003228.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Bloc1s1

          Panther Classification:

          Molecular Function -

          membrane traffic protein

          Gene Ontology Categories:

          Function(s) Process(es)

          anterograde axonal transport
          aerobic respiration
          peptidyl-lysine acetylation
          neuron projection development
          anterograde synaptic vesicle transport
          mitophagy in response to mitochondrial depolarization
          protein binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27543923 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27543925 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1992650 AK003035.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1992650 BC034662.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1992650 NM_015740.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 242804 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 18369 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P137023 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1992650 AK003035.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1992650 BC034662.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1992650 NM_015740.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2110027 - -
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P142113 AK003035.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P142113 BC034662.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P142113 NM_015740.3 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27543898 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27543949 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27543968 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27543930 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27543928 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27543903 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27543924 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27543919 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27543969 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27543947 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27543936 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27543914 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27543954 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27543942 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27543944 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27543916 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27543973 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27543922 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27543907 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27543921 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27543902 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27543965 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27543976 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27543933 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27543963 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27543945 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27543931 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27543910 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27543920 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27543960 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27543917 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27543978 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27543938 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27543952 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27543951 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27543971 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27543940 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27543958 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27543979 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27543974 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27543957 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P137023 - -
          Whole Mouse Genome G4122A A_51_P142113 AK003035.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P142113 BC034662.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P142113 NM_015740.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P546540 AK003035.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P546540 BC034662.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P546540 NM_015740.3 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P124391 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P137023 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08692362 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08692367 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08692369 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08744351 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08273301 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08744346 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08612293 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08692358 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08273302 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08273290 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27543923 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27543925 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543979 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543880 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543896 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543969 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543920 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543934 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543944 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543865 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543834 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543841 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543911 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543949 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543929 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543831 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543849 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543957 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543951 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543978 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543916 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543843 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543963 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543825 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543894 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543827 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543890 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543836 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543839 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543954 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543973 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543936 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543861 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543914 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543921 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543917 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543872 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543931 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543933 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543833 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543857 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543874 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543885 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543947 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543878 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543853 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543910 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543883 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543945 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543968 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543901 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543907 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543873 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543903 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543961 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543864 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543876 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543829 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543855 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543854 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543859 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543924 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543976 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27543965 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08665346 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08209070 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27543902 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27543936 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27543933 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27543939 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27543907 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27543898 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27543911 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27543903 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08692359 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27543945 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27543905 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27543938 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27543924 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27543901 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27543922 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27543910 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27543920 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27543941 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27543934 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27543916 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27543942 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27543917 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27543929 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27543921 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27543940 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10457925 - -
          Affymetrix Mouse Gene 1.0 ST Array 10373598 - -
          Affymetrix Mouse Gene 1.0 ST Array 10373597 - -
          Affymetrix Mouse Gene 1.0 ST Array 10373595 - -
          Affymetrix Mouse Gene 1.0 ST Array 10373596 - -
          Affymetrix Mouse Gene 1.0 ST Array 10457926 - -
          Affymetrix Mouse Gene 1.0 ST Array 10373599 - -
          Affymetrix HT MG-430 PM Array Plate 1422614_PM_s_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10373594 - -
          Affymetrix Mouse Gene 1.1 ST Array 10457924 - -
          Mouse Genome 430 2.0 Array 1422614_PM_s_at - -
          Mouse Genome 430 2.0 Array 1422614_s_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 14533_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 14533_at - -
          Affymetrix Mouse Clariom S Array PSR1000024058.mm.1 NM_015740.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000024058.mm.1 NM_015740.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1000024058.mm.1 TC1000003228.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000023916.mm.1 NM_015740.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000023916.mm.1 NM_015740.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1000023916.mm.1 TC1000003228.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000023924.mm.1 NM_015740.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000023924.mm.1 NM_015740.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1000023924.mm.1 TC1000003228.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000023918.mm.1 NM_015740.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000023918.mm.1 NM_015740.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1000023918.mm.1 TC1000003228.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000023920.mm.1 NM_015740.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000023920.mm.1 NM_015740.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1000023920.mm.1 TC1000003228.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000023917.mm.1 NM_015740.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000023917.mm.1 NM_015740.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1000023917.mm.1 TC1000003228.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000023913.mm.1 NM_015740.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000023913.mm.1 NM_015740.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1000023913.mm.1 TC1000003228.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000023925.mm.1 NM_015740.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000023925.mm.1 NM_015740.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1000023925.mm.1 TC1000003228.mm.1 MTA TC
          Mouse Expression Set 430 1422614_PM_s_at - -
          Mouse Expression Set 430 1422614_s_at - -
          Murine Genome U74v2 Set 97917_PM_at - -
          Murine Genome U74v2 Set 97917_at - -
          Mouse Genome 430A 2.0 Array 1422614_s_at - -
          Mouse Genome 430A 2.0 Array 1422614_PM_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1422614_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1422614_PM_s_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 3940717 - -
          Illumina mouse-6 v1.0 expression beadchip 130341 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000130341 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003940717 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2649256 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2726630 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2726630 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2649256 - -
          mouse-6 v1.1 (Illumina) scl37341.3.1_8-S - -
          mouse-6 v1.1 (Illumina) scl0003930.1_2-S - -
          Illumina Sentrix mouse-6.v1 bead array 130341 - -
          Illumina Sentrix mouse-6.v1 bead array 3940717 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0003930.1_2-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl37341.3.1_8-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3940717 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 130341 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1235757 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1235757 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0003930.1_2-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl37341.3.1_8-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline