Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00497362_m1
          See other CRLF2 GE Assays ›
          Gene Symbol:
          Crlf2
          Gene Name:
          cytokine receptor-like factor 2
          Gene Aliases:
          CRLM2, Ly114, Tpte2, Tslpr
          Chromosome Location:
          Chr.5: 109554709 - 109559017 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00497362_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Crlf2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001164735.1 NP_001158207.1 3 - 4 453 146
          NM_016715.4 NP_057924.3 3 - 4 453 146
          GenBank mRNA AB031333.1 - 3 - 4 401 146
          AB039945.1 - 3 - 4 349 146
          AF201963.1 - 3 - 4 504 146
          AF232936.1 - 3 - 4 417 146
          AK149928.1 - 3 - 4 454 146
          BC016580.1 - 3 - 4 531 2655857 146
          BC023788.1 - 3 - 4 441 5346748 146

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500002969.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Crlf2

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          immunoglobulin secretion involved in immune response
          inflammatory response
          T-helper 2 cell cytokine production
          cytokine activity

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0500026287.mm.1 TC0500002969.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500026287.mm.1 NM_001164735.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500026287.mm.1 NM_001164735.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500026287.mm.1 NM_016715.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500026287.mm.1 NM_016715.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0500026285.mm.1 TC0500002969.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500026285.mm.1 NM_001164735.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500026285.mm.1 NM_001164735.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500026285.mm.1 NM_016715.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500026285.mm.1 NM_016715.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0500026282.mm.1 TC0500002969.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500026282.mm.1 NM_001164735.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500026282.mm.1 NM_001164735.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500026282.mm.1 NM_016715.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500026282.mm.1 NM_016715.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0500026283.mm.1 TC0500002969.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500026283.mm.1 NM_001164735.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500026283.mm.1 NM_001164735.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500026283.mm.1 NM_016715.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500026283.mm.1 NM_016715.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0500026281.mm.1 TC0500002969.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500026281.mm.1 NM_001164735.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500026281.mm.1 NM_001164735.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500026281.mm.1 NM_016715.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500026281.mm.1 NM_016715.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0500026284.mm.1 TC0500002969.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 455897 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline