| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0700007560.mm.1 |
TC0700000861.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700007560.mm.1 |
NM_016974.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700007560.mm.1 |
NM_016974.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700007563.mm.1 |
TC0700000861.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700007563.mm.1 |
NM_016974.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700007563.mm.1 |
NM_016974.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700007559.mm.1 |
TC0700000861.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700007559.mm.1 |
NM_016974.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700007559.mm.1 |
NM_016974.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700007562.mm.1 |
TC0700000861.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700007562.mm.1 |
NM_016974.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700007562.mm.1 |
NM_016974.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700007561.mm.1 |
TC0700000861.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700007561.mm.1 |
NM_016974.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700007561.mm.1 |
NM_016974.3 |
Refseq |