Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00498108_m1
          See other ZFP113 GE Assays ›
          Gene Symbol:
          Zfp113
          Gene Name:
          zinc finger protein 113
          Gene Aliases:
          4732456B05Rik, mKIAA4229
          Chromosome Location:
          Chr.5: 138139702 - 138155744 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00498108_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zfp113
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_019747.4 NP_062721.2 4 - 5 466 87
          GenBank mRNA AK076349.1 - 4 - 5 461 87
          AK220556.1 - 4 - 5 466 87
          BC052453.1 - 4 - 5 410 6410632 87
          BC054525.1 - 4 - 5 410 6410632 87
          BC056445.1 - 3 - 4 331 5703904 87
          BC056991.1 - 4 - 5 413 87

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500003470.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Zfp113

          Panther Classification:

          Molecular Function -

          C2H2 zinc finger transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          identical protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24129054 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24129051 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2010008 CJ244868.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2010008 NM_019747.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2010003 AK081430.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2010003 AK220556.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2010003 BC052453.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2010003 BC054525.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2010003 NM_019747.4 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 847280 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 467615 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 504716 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 736784 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 531573 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 870954 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 555358 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 199059 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 366450 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 589923 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 167564 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P117778 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P113524 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2010008 CJ244868.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2010008 NM_019747.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2010003 AK081430.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2010003 AK220556.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2010003 BC052453.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2010003 BC054525.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2010003 NM_019747.4 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24129045 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24129051 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24129074 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24129069 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24129063 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24129054 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24129056 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24129055 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24129068 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24129061 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24129066 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24129042 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24129044 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24129057 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24129071 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24129060 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24129049 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P113524 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05762296 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05762274 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P117778 - -
          Whole Mouse Genome G4122A A_52_P507890 AK081430.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P507890 BC052453.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P507890 BC054525.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P507890 NM_019747.4 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P103475 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P117778 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P113524 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04535318 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04684838 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04535315 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04293286 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04292238 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24129051 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24129054 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24128951 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24128977 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24128987 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129016 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129055 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129040 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129107 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129014 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129032 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24128948 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24128965 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24128979 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129044 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129045 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129057 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129010 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129079 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129084 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24128968 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129033 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129111 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24128961 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129051 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24128997 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129063 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24128999 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129049 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129003 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129012 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24128983 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24128959 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24128954 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24128993 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129088 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24128958 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24128953 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24128988 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24128969 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24128975 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24128984 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129060 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129019 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129092 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24128963 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129069 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129061 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24128998 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129075 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129089 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24128970 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24128964 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129009 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24128950 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129090 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129035 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129070 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129066 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129085 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129105 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129109 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129072 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129077 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129002 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24128985 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129081 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129007 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129006 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24128972 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129068 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129074 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24128957 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24129020 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24129028 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24129042 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24129030 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04169525 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24129057 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24129049 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24129035 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24129045 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24129055 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24129063 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24129029 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24129019 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24129031 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24129037 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24129058 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24129051 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24129039 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24129040 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24129033 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24129032 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04169526 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24129043 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24129044 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24129061 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10534968 - -
          Affymetrix Mouse Gene 1.0 ST Array 10534970 - -
          Affymetrix Mouse Gene 1.0 ST Array 10534972 - -
          Affymetrix Mouse Gene 1.0 ST Array 10534971 - -
          Affymetrix Mouse Gene 1.0 ST Array 10534969 - -
          Affymetrix Mouse Gene 1.0 ST Array 10534967 - -
          Affymetrix Mouse Gene 1.0 ST Array 10534973 - -
          Affymetrix HT MG-430 PM Array Plate 1420675_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10534966 - -
          Mouse Genome 430 2.0 Array 1420675_PM_at - -
          Mouse Genome 430 2.0 Array 1420675_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 56314_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 56314_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0500031620.mm.1 TC0500003470.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500031620.mm.1 NM_019747.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500031620.mm.1 NM_019747.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0500031618.mm.1 TC0500003470.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500031618.mm.1 NM_019747.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500031618.mm.1 NM_019747.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0500031621.mm.1 TC0500003470.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500031621.mm.1 NM_019747.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500031621.mm.1 NM_019747.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0500031623.mm.1 TC0500003470.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500031623.mm.1 NM_019747.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500031623.mm.1 NM_019747.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0500031622.mm.1 TC0500003470.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500031622.mm.1 NM_019747.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500031622.mm.1 NM_019747.4 Refseq
          Mouse Expression Set 430 1420675_PM_at - -
          Mouse Expression Set 430 1420675_at - -
          Murine Genome U74v2 Set 129141_PM_at - -
          Murine Genome U74v2 Set 129141_at - -
          Mouse Genome 430A 2.0 Array 1420675_at - -
          Mouse Genome 430A 2.0 Array 1420675_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1420675_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1420675_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 542663 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2320047 - -
          MouseWG-6 v2 Expression BeadChip 1770193 - -
          Illumina mouse-6 v1.0 expression beadchip 2570273 - -
          Illumina mouse-6 v1.0 expression beadchip 2320047 - -
          Illumina mouse-6 v1.0 expression beadchip 1770193 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001770193 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002570273 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002320047 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2427924 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2439322 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2423988 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2427924 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2439322 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2423988 - -
          mouse-6 v1.1 (Illumina) scl0056314.2_187-S - -
          mouse-6 v1.1 (Illumina) scl0004028.1_1-S - -
          mouse-6 v1.1 (Illumina) scl056314.1_96-S - -
          Illumina Sentrix mouse-6.v1 bead array 2570273 - -
          Illumina Sentrix mouse-6.v1 bead array 1770193 - -
          Illumina Sentrix mouse-6.v1 bead array 2320047 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0004028.1_1-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0056314.2_187-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl056314.1_96-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2570273 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1770193 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2320047 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2439322 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2423988 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2439322 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2423988 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline