Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00498219_m1
          See other VMN2R26 GE Assays ›
          Gene Symbol:
          Vmn2r26
          Gene Name:
          vomeronasal 2, receptor 26
          Gene Aliases:
          V2R1, V2r1b
          Chromosome Location:
          Chr.6: 124024758 - 124062035 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00498219_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Vmn2r26
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_019917.2 NP_064301.2 3 - 4 1321 112
          GenBank mRNA AF053985.1 - 3 - 4 1321 112
          AY426342.1 - 3 - 4 1321 112

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600001414.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Vmn2r26

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          signal transduction
          G-protein coupled receptor signaling pathway
          response to pheromone
          signal transducer activity
          G-protein coupled receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P103474 AY426342.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P103474 NM_019917.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P103474 AF053985.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 772857 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 560249 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 176630 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 165377 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P142382 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P103474 AY426342.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P103474 NM_019917.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P103474 AF053985.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P100527 AF053985.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24813719 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24813718 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24813741 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06054757 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06054769 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P142382 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01436899 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06054749 - -
          Whole Mouse Genome G4122A A_51_P100527 AF053985.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P103474 AY426342.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P103474 NM_019917.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P103474 AF053985.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P126282 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P142382 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P06054749 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P01436899 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24813862 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24813824 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24813823 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24813827 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24813773 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24813818 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24813868 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24813855 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24813829 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24813863 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24813826 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24813821 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24813854 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24813769 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24813828 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24813743 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24813719 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24813741 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10547690 - -
          Affymetrix Mouse Gene 1.0 ST Array 10541637 - -
          Affymetrix Mouse Gene 1.0 ST Array 10541639 - -
          Affymetrix Mouse Gene 1.0 ST Array 10541628 - -
          Affymetrix Mouse Gene 1.0 ST Array 10541638 - -
          Affymetrix Mouse Gene 1.0 ST Array 10541640 - -
          Affymetrix Mouse Gene 1.0 ST Array 10547675 - -
          Affymetrix Mouse Gene 1.0 ST Array 10541626 - -
          Affymetrix Mouse Gene 1.0 ST Array 10541641 - -
          Affymetrix HT MG-430 PM Array Plate 1421719_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10541636 - -
          Affymetrix Mouse Gene 1.1 ST Array 10547689 - -
          Affymetrix Mouse Gene 1.1 ST Array 10547674 - -
          Affymetrix Mouse Gene 1.1 ST Array 10541625 - -
          Affymetrix Mouse Gene 1.1 ST Array 10541627 - -
          Mouse Genome 430 2.0 Array 1421719_at - -
          Mouse Genome 430 2.0 Array 1421719_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0600011369.mm.1 TC0600001414.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600011369.mm.1 NM_019917.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600011369.mm.1 NM_019917.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0600011372.mm.1 TC0600001414.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600011372.mm.1 NM_019917.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600011372.mm.1 NM_019917.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0600011370.mm.1 TC0600001414.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600011370.mm.1 NM_019917.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600011370.mm.1 NM_019917.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0600011371.mm.1 TC0600001414.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600011371.mm.1 NM_019917.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600011371.mm.1 NM_019917.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0600011373.mm.1 TC0600001414.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600011373.mm.1 NM_019917.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600011373.mm.1 NM_019917.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0600011374.mm.1 TC0600001414.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600011374.mm.1 NM_019917.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600011374.mm.1 NM_019917.2 Refseq
          Mouse Expression Set 430 1421719_at - -
          Mouse Expression Set 430 1421719_PM_at - -
          Murine Genome U74v2 Set 99700_PM_at - -
          Murine Genome U74v2 Set 99700_at - -
          Mouse Genome 430A 2.0 Array 1421719_PM_at - -
          Mouse Genome 430A 2.0 Array 1421719_at - -
          Affymetrix HT Mouse Genome 430A Array 1421719_at - -
          Affymetrix HT Mouse Genome 430A Array 1421719_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 598109 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6770176 - -
          Illumina mouse-6 v1.0 expression beadchip 6770176 - -
          Illumina mouse-6 v1.0 expression beadchip 2900332 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006770176 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002900332 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1245899 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2473322 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2473322 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1245899 - -
          mouse-6 v1.1 (Illumina) scl056552.6_38-S - -
          mouse-6 v1.1 (Illumina) scl056552.3_195-S - -
          Illumina Sentrix mouse-6.v1 bead array 6770176 - -
          Illumina Sentrix mouse-6.v1 bead array 2900332 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl056552.3_195-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl056552.6_38-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2900332 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6770176 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2473322 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2473327 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline