Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00498295_m1
          See other UBE2L6 GE Assays ›
          Gene Symbol:
          Ube2l6
          Gene Name:
          ubiquitin-conjugating enzyme E2L 6
          Gene Aliases:
          2810489I21Rik, RIG-B, UBCH8, Ubce8, Ubcm8
          Chromosome Location:
          Chr.2: 84798828 - 84810003 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00498295_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ube2l6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_019949.2 NP_064333.2 2 - 3 198 92
          GenBank mRNA AF159230.1 - 2 - 3 184 92
          AK010942.1 - 2 - 3 177 92
          AK013452.1 - 2 - 3 188 92
          AK152010.1 - 2 - 3 178 92
          AK152066.1 - 2 - 3 151 92
          AK152113.1 - 2 - 3 199 92
          AK168695.1 - 2 - 3 136 92
          AK171629.1 - 2 - 3 147 92
          BC008238.1 - 2 - 3 166 2651702 92
          CT010270.1 - 2 - 3 117 92

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200001222.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ube2l6

          Panther Classification:

          Molecular Function -

          ubiquitin-protein ligase

          Gene Ontology Categories:

          Function(s) Process(es)

          modification-dependent protein catabolic process
          ISG15-protein conjugation
          nucleotide binding
          protein binding
          ATP binding
          ligase activity
          ubiquitin-like protein transferase activity
          ISG15 transferase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2141943 AK013452.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2031125 CJ300190.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2031125 AK152113.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2031125 AK168695.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2031125 AK171629.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2031125 AK206200.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 263010 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 456677 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 549566 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 671590 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 635543 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 937471 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 170652 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 27722 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P174611 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P111022 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2031125 CJ300190.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2031125 AK152113.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2031125 AK168695.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2031125 AK171629.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2031125 AK206200.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2141943 AK013452.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P214740 AK152113.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P214740 AK168695.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P214740 AK171629.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P214740 AK212910.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P214740 BC008238.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21394642 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21394656 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21394646 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21394657 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21394645 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21394628 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21394652 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21394626 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21394640 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21394653 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21394648 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21394638 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21394637 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21394654 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21394643 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21394650 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P111022 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P174611 - -
          Whole Mouse Genome G4122A A_52_P214740 AK152113.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P214740 AK168695.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P214740 AK171629.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P214740 AK212910.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P214740 BC008238.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P106566 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P174611 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P111022 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01413663 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01413661 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01413655 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01413657 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394647 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394609 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394613 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394751 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394756 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394621 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394705 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394682 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394741 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394619 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394761 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394612 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394725 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394769 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394763 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394603 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394677 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394602 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394640 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394606 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394726 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394735 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394645 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394688 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394727 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394667 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394744 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394758 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394671 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394728 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394768 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394714 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394672 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394700 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394739 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394719 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394721 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394646 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394652 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394664 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394729 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394670 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394623 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394755 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394747 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394656 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394733 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394723 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394648 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394650 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394692 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394766 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394673 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394643 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394712 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394615 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394753 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21394750 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21394602 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01389795 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21394640 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21394680 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21394606 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01590154 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21394664 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01614465 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02187072 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21394684 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21394667 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21394673 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21394687 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21394603 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21394623 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21394676 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21394670 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21394648 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01413663 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01172948 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21394619 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21394646 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21394628 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21394647 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21394677 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01349790 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02192479 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21394652 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21394624 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21394645 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21394662 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21394621 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21394650 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21394626 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21394653 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02214468 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21394682 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21394675 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21394618 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21394638 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21394643 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01239053 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21394617 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10473358 - -
          Affymetrix Mouse Gene 1.0 ST Array 10473361 - -
          Affymetrix Mouse Gene 1.0 ST Array 10473360 - -
          Affymetrix Mouse Gene 1.0 ST Array 10473357 - -
          Affymetrix Mouse Gene 1.0 ST Array 10473359 - -
          Affymetrix HT MG-430 PM Array Plate 1417172_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10473356 - -
          Mouse Genome 430 2.0 Array 1417172_PM_at - -
          Mouse Genome 430 2.0 Array 1417172_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 56791_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 56791_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0200009748.mm.1 NM_019949.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200009748.mm.1 NM_019949.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200009748.mm.1 TC0200001222.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200009750.mm.1 NM_019949.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200009750.mm.1 NM_019949.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200009750.mm.1 TC0200001222.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200009746.mm.1 NM_019949.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200009746.mm.1 NM_019949.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200009746.mm.1 TC0200001222.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200009747.mm.1 NM_019949.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200009747.mm.1 NM_019949.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200009747.mm.1 TC0200001222.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200009749.mm.1 NM_019949.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200009749.mm.1 NM_019949.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200009749.mm.1 TC0200001222.mm.1 MTA TC
          Mouse Expression Set 430 1417172_at - -
          Mouse Expression Set 430 1417172_PM_at - -
          Murine Genome U74v2 Set 98597_at - -
          Murine Genome U74v2 Set 98597_PM_at - -
          Mouse Genome 430A 2.0 Array 1417172_PM_at - -
          Mouse Genome 430A 2.0 Array 1417172_at - -
          Affymetrix HT Mouse Genome 430A Array 1417172_at - -
          Affymetrix HT Mouse Genome 430A Array 1417172_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 401185 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2680427 - -
          Illumina mouse-6 v1.0 expression beadchip 2680427 - -
          Illumina mouse-6 v1.0 expression beadchip 1170687 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002680427 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001170687 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2431619 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2486267 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2486267 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2431619 - -
          mouse-6 v1.1 (Illumina) scl056791.4_30-S - -
          mouse-6 v1.1 (Illumina) scl0003139.1_11-S - -
          Illumina Sentrix mouse-6.v1 bead array 1170687 - -
          Illumina Sentrix mouse-6.v1 bead array 2680427 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl056791.4_30-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0003139.1_11-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2680427 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1170687 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2486267 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2486267 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2431619 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0003139.1_11-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline