Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00498358_m1
          See other PSRC1 GE Assays ›
          Gene Symbol:
          Psrc1
          Gene Name:
          proline/serine-rich coiled-coil 1
          Gene Aliases:
          5430413I02Rik, AL033312, Dda3
          Chromosome Location:
          Chr.3: 108383804 - 108388231 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00498358_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Psrc1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001190161.1 NP_001177090.1 4 - 5 640 65
          NM_019976.3 NP_064360.1 4 - 5 637 65
          GenBank mRNA AF156598.1 - 4 - 5 632 65
          AK011197.1 - 4 - 5 612 65
          AK017302.1 - 4 - 5 579 65
          AK041835.1 - 4 - 5 587 65
          AK167415.1 - 4 - 5 583 65
          BC019165.1 - 4 - 5 595 3963608 65

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0300001142.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Psrc1

          Gene Ontology Categories:

          Function(s) Process(es)

          microtubule bundle formation
          cell cycle
          mitotic nuclear division
          mitotic metaphase plate congression
          cellular process
          negative regulation of cell growth
          positive regulation of microtubule polymerization
          positive regulation of cyclin-dependent protein serine/threonine kinase activity
          positive regulation of transcription, DNA-templated
          cell division
          regulation of mitotic spindle organization
          protein binding
          microtubule binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22412278 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22412275 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22412277 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22412279 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22412276 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2429225 BC019165.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2429225 AK017302.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2429225 AK041835.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2429225 AK167415.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2429225 NM_019976.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 47630 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 513286 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2429225 BC019165.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2429225 AK017302.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2429225 AK041835.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2429225 AK167415.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2429225 NM_019976.3 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22412275 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22412259 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22412273 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22412254 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22412291 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22412284 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22412262 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22412287 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22412256 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22412296 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22412265 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22412281 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22412286 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22412271 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22412263 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22412295 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22412294 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22412276 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22412289 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22412290 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22412282 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22412261 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22412278 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22412257 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22412268 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22412269 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22412298 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22412280 - -
          Whole Mouse Genome G4122A A_51_P409694 BC019165.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P409694 AK041835.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P409694 AK167415.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P409694 NM_019976.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P409694 AF156598.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P493112 BC008994.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P493112 BC019165.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P493112 AK017302.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P493112 AK041835.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P493112 AK167415.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P271528 BC019165.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P271528 AK017302.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P271528 AK041835.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P271528 AK167415.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P271528 NM_019976.3 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P123515 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02364886 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02364881 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02324133 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02364888 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02364878 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02364880 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02364882 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22412278 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22412279 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22412275 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22412277 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22412276 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22412255 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22412282 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22412319 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22412256 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22412298 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22412285 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22412299 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22412289 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22412268 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22412261 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22412310 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22412306 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22412314 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22412316 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22412258 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22412259 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22412265 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22412281 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22412304 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22412273 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22412295 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22412278 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22412286 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22412303 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22412254 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22412305 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22412271 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22412287 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22412276 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22412284 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22412313 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22412262 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22412301 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22412276 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22412273 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22412271 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22412304 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22412305 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22412262 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22412299 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02895025 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02364886 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22412313 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02324140 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22412291 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02981157 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02444829 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22412281 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22412296 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22412286 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22412316 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02444832 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22412301 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22412282 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22412261 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22412287 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22412265 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22412280 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02611202 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22412266 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22412285 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22412278 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03133789 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22412259 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22412297 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22412306 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22412263 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02364893 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03133785 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22412310 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22412269 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22412303 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02611201 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10495318 - -
          Affymetrix Mouse Gene 1.0 ST Array 10495317 - -
          Affymetrix Mouse Gene 1.0 ST Array 10495319 - -
          Affymetrix Mouse Gene 1.0 ST Array 10495322 - -
          Affymetrix Mouse Gene 1.0 ST Array 10495329 - -
          Affymetrix Mouse Gene 1.0 ST Array 10495328 - -
          Affymetrix Mouse Gene 1.0 ST Array 10495320 - -
          Affymetrix Mouse Gene 1.0 ST Array 10495326 - -
          Affymetrix Mouse Gene 1.0 ST Array 10495324 - -
          Affymetrix HT MG-430 PM Array Plate 1417323_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1451698_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1425416_PM_s_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10495316 - -
          Mouse Genome 430 2.0 Array 1425416_PM_s_at - -
          Mouse Genome 430 2.0 Array 1451698_PM_at - -
          Mouse Genome 430 2.0 Array 1417323_PM_at - -
          Mouse Genome 430 2.0 Array 1451698_at - -
          Mouse Genome 430 2.0 Array 1425416_s_at - -
          Mouse Genome 430 2.0 Array 1417323_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 56742_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 56742_at - -
          Affymetrix Mouse Clariom S Array PSR0300009311.mm.1 NM_001190161.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300009311.mm.1 NM_001190161.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300009311.mm.1 NM_019976.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300009311.mm.1 NM_019976.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300009311.mm.1 TC0300001142.mm.1 MTA TC
          Mouse Expression Set 430 1417323_PM_at - -
          Mouse Expression Set 430 1417323_at - -
          Mouse Genome 430A 2.0 Array 1417323_PM_at - -
          Mouse Genome 430A 2.0 Array 1417323_at - -
          Affymetrix HT Mouse Genome 430A Array 1451698_at - -
          Affymetrix HT Mouse Genome 430A Array 1425416_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1417323_at - -
          Affymetrix HT Mouse Genome 430A Array 1451698_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1425416_PM_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1417323_PM_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 5360154 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005360154 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2692441 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2692441 - -
          mouse-6 v1.1 (Illumina) scl22720.6.1_30-S - -
          Illumina Sentrix mouse-6.v1 bead array 5360154 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl22720.6.1_30-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5360154 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2692441 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2692441 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl22720.6.1_30-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline