Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00498365_m1
          See other TEX101 GE Assays ›
          Gene Symbol:
          Tex101
          Gene Name:
          testis expressed gene 101
          Gene Aliases:
          1700008H15Rik, AI429076, TES101RP
          Chromosome Location:
          Chr.7: 24668012 - 24672050 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00498365_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tex101
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_019981.2 NP_064365.1 4 - 5 531 71
          GenBank mRNA AB022914.1 - 4 - 5 529 71
          AK005769.1 - 4 - 5 525 71
          BC048475.1 - 4 - 5 522 6740387 71
          DQ222038.1 - 730 71
          DQ222039.1 - 668 71
          DQ222040.1 - 1031 71
          DQ222041.1 - 972 71
          DQ222042.1 - 4 - 5 533 71

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700002609.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tex101

          Gene Ontology Categories:

          Function(s)

          positive regulation of leukocyte activation
          positive regulation of peptidyl-tyrosine phosphorylation
          positive regulation of release of sequestered calcium ion into cytosol

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P499158 AB022914.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P499158 AK005769.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P499158 BC048475.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P499158 NM_019981.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P499158 AB022914.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P499158 AK005769.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P499158 BC048475.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P499158 NM_019981.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 811180 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 139163 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 77852 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 330225 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 105865 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P139004 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P499158 AB022914.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P499158 AK005769.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P499158 BC048475.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P499158 NM_019981.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P499158 AB022914.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P499158 AK005769.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P499158 BC048475.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P499158 NM_019981.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25006092 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25006090 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25006080 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25006115 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25006119 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25006071 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25006075 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25006076 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25006072 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25006073 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25006086 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25006074 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25006070 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25006088 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25006067 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25006117 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25006065 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25006083 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25006089 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25006085 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25006098 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25006097 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25006094 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25006095 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25006068 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P139004 - -
          Whole Mouse Genome G4122A A_52_P499158 AB022914.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P499158 AK005769.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P499158 BC048475.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P499158 NM_019981.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P499158 AB022914.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P499158 AK005769.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P499158 BC048475.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P499158 NM_019981.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P132792 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P102442 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P139004 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06664663 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06252003 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06664648 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06664649 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006024 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006018 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006136 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006050 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006067 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006051 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006133 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006080 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006047 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006059 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006061 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006120 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006085 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006062 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006071 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006124 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006117 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006054 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006083 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006040 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006070 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006139 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006076 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006029 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006074 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006035 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006113 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006038 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006088 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006033 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006090 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006030 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006063 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006128 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006094 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006127 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006057 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006073 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006129 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006130 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006052 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006065 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006115 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006097 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006126 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006086 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006135 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006019 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25006021 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05985788 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25006119 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25006097 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25006115 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25006054 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25006052 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25006069 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25006080 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25006136 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25006098 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06664668 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25006071 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25006139 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25006065 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25006133 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25006094 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25006085 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25006095 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25006120 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06152574 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25006059 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25006135 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06314821 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25006074 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25006057 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06196302 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25006092 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06664670 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25006061 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25006062 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25006117 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25006068 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25006128 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06515526 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25006086 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25006050 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25006067 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06664644 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06313286 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06434602 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06664663 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06664657 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25006113 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25006048 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25006073 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06196298 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10560878 - -
          Affymetrix Mouse Gene 1.0 ST Array 10560879 - -
          Affymetrix Mouse Gene 1.0 ST Array 10560875 - -
          Affymetrix Mouse Gene 1.0 ST Array 10560876 - -
          Affymetrix Mouse Gene 1.0 ST Array 10560877 - -
          Affymetrix Mouse Gene 1.0 ST Array 10560874 - -
          Affymetrix HT MG-430 PM Array Plate 1449103_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10560873 - -
          Mouse Genome 430 2.0 Array 1449103_PM_at - -
          Mouse Genome 430 2.0 Array 1449103_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 56746_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 56746_at - -
          Affymetrix Mouse Clariom S Array PSR0700022492.mm.1 TC0700002609.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700022492.mm.1 NM_019981.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700022492.mm.1 NM_019981.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700022493.mm.1 TC0700002609.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700022493.mm.1 NM_019981.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700022493.mm.1 NM_019981.2 Refseq
          Mouse Expression Set 430 1449103_at - -
          Mouse Expression Set 430 1449103_PM_at - -
          Mouse Genome 430A 2.0 Array 1449103_at - -
          Mouse Genome 430A 2.0 Array 1449103_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1449103_at - -
          Affymetrix HT Mouse Genome 430A Array 1449103_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 676603 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 540133 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000540133 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2590670 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2590670 - -
          mouse-6 v1.1 (Illumina) scl31647.6.1_68-S - -
          Illumina Sentrix mouse-6.v1 bead array 540133 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl31647.6.1_68-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 540133 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2590670 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2590670 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline