Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00498531_m1
          See other DNAJC4 GE Assays ›
          Gene Symbol:
          Dnajc4
          Gene Name:
          DnaJ (Hsp40) homolog, subfamily C, member 4
          Gene Aliases:
          2010301J22Rik, Hspf2, Mcg18
          Chromosome Location:
          Chr.19: 6987911 - 6992696 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00498531_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dnajc4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_020566.1 NP_065591.1 1 - 2 120 77
          GenBank mRNA AF036875.1 - 1 - 2 120 77
          AK170008.1 - 1 - 2 359 77
          BC049281.1 - 1 - 2 351 4989485 77

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1900001028.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Dnajc4

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31937161 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31937160 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P231738 BC049281.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P231738 NM_020566.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P231738 AF036875.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P231738 AK008507.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P231738 AK170008.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P231738 BC049281.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P231738 NM_020566.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P231738 AF036875.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P231738 AK008507.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P231738 AK170008.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 566970 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 333134 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 866107 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 174008 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P151491 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P231738 BC049281.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P231738 NM_020566.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P231738 AF036875.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P231738 AK008507.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P231738 AK170008.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P231738 BC049281.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P231738 NM_020566.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P231738 AF036875.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P231738 AK008507.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P231738 AK170008.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P231738 BC049281.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P231738 NM_020566.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P231738 AF036875.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P231738 AK008507.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P231738 AK170008.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P231738 BC049281.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P231738 NM_020566.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P231738 AF036875.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P231738 AK008507.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P231738 AK170008.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31937167 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31937190 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31937182 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31937189 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31937168 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31937187 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31937174 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31937186 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31937183 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31937160 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31937163 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31937178 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31937177 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31937180 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31937176 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31937165 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31937194 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31937171 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31937191 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P151491 - -
          Whole Mouse Genome G4122A A_51_P231738 BC049281.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P231738 NM_020566.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P231738 AF036875.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P231738 AK008507.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P231738 AK170008.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P231738 BC049281.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P231738 NM_020566.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P231738 AF036875.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P231738 AK008507.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P231738 AK170008.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P104809 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P151491 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14267109 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14267112 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14489804 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14489812 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14489820 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14267113 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31937161 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31937160 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937220 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937090 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937168 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937134 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937140 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937150 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937227 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937221 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937152 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937234 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937128 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937095 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937171 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937121 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937211 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937135 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937236 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937131 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937176 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937241 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937115 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937165 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937125 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937148 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937151 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937178 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937143 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937240 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937174 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937232 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937183 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937163 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937189 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937158 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937103 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937194 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937112 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937106 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937187 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937218 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937222 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937099 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937180 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937238 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937155 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937219 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937119 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937097 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937216 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937225 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937091 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937108 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937093 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937190 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937209 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937098 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937101 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937223 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937230 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937126 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937184 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937167 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937177 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937137 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937228 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937156 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937213 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937105 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937160 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14176385 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31937184 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31937178 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14267116 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31937190 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31937163 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14383123 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31937160 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31937187 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31937165 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31937186 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31937180 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31937174 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31937171 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31937167 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31937177 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31937194 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10465574 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465576 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465578 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465572 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465573 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465575 - -
          Affymetrix Mouse Gene 1.0 ST Array 10465579 - -
          Affymetrix HT MG-430 PM Array Plate 1417934_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10465571 - -
          Mouse Genome 430 2.0 Array 1417934_PM_at - -
          Mouse Genome 430 2.0 Array 1417934_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 57431_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 57431_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1900009584.mm.1 TC1900001028.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900009582.mm.1 NM_020566.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900009582.mm.1 NM_020566.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900009582.mm.1 TC1900001028.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900009583.mm.1 NM_020566.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900009583.mm.1 NM_020566.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900009583.mm.1 TC1900001028.mm.1 MTA TC
          Mouse Expression Set 430 1417934_PM_at - -
          Mouse Expression Set 430 1417934_at - -
          Murine Genome U74v2 Set 102324_PM_at - -
          Murine Genome U74v2 Set 102324_at - -
          Mouse Genome 430A 2.0 Array 1417934_PM_at - -
          Mouse Genome 430A 2.0 Array 1417934_at - -
          Affymetrix HT Mouse Genome 430A Array 1417934_at - -
          Affymetrix HT Mouse Genome 430A Array 1417934_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 807241 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 4850136 - -
          Illumina mouse-6 v1.0 expression beadchip 4850136 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004850136 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2646976 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2646976 - -
          mouse-6 v1.1 (Illumina) scl52898.8.1_85-S - -
          Illumina Sentrix mouse-6.v1 bead array 4850136 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl52898.8.1_85-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4850136 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2646976 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2646976 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl52898.8.1_85-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline