Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00498614_m1
          See other SLC46A2 GE Assays ›
          Gene Symbol:
          Slc46a2
          Gene Name:
          solute carrier family 46, member 2
          Gene Aliases:
          5430429N04Rik, Ly110, TSO-1C12, Tscot
          Chromosome Location:
          Chr.4: 59905899 - 59915056 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00498614_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Slc46a2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_021053.4 NP_066395.3 3 - 4 1513 64
          GenBank mRNA AF148145.1 - 3 - 4 1485 64
          AK017363.1 - 3 - 4 1513 64
          BC140962.1 - 3 - 4 1446 9055574 64

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400002765.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Slc46a2

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          T cell homeostasis
          regulation of T cell differentiation
          thymus development
          transmembrane transport
          negative regulation of T cell apoptotic process
          molecular_function
          transporter activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P185763 AK017363.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P185763 AK040063.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P185763 NM_021053.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P185763 AF148145.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P185763 AK017363.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P185763 AK040063.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P185763 NM_021053.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P185763 AF148145.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 117624 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 453067 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 875729 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 8745 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 62671 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P107885 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P185763 AK017363.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P185763 AK040063.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P185763 NM_021053.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P185763 AF148145.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P185763 AK017363.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P185763 AK040063.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P185763 NM_021053.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P185763 AF148145.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P185763 AK017363.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P185763 AK040063.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P185763 NM_021053.4 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P185763 AF148145.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P185763 AK017363.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P185763 AK040063.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P185763 NM_021053.4 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P185763 AF148145.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22959043 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22959024 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22959068 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22959044 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22959033 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22959069 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22959062 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22959028 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22959046 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22959066 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22959035 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22959042 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22959055 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22959052 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22959060 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22959025 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22959061 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22959032 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22959029 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22959036 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22959056 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22959067 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22959048 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22959040 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22959049 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22959030 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22959065 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22959054 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22959058 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22959047 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22959070 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22959037 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P107885 - -
          Whole Mouse Genome G4122A A_51_P185763 AK017363.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P185763 AK040063.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P185763 NM_021053.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P185763 AF148145.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P185763 AK017363.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P185763 AK040063.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P185763 NM_021053.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P185763 AF148145.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P127729 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P107885 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03963126 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03258609 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03258610 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03930566 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03258611 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03963122 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03258598 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03258613 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22959049 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22958998 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22959068 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22959013 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22959032 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22959067 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22958996 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22958986 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22959028 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22958987 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22959024 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22959017 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22959029 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22958991 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22959044 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22959086 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22959035 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22959052 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22959063 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22959072 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22959061 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22959055 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22959037 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22959019 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22959065 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22959070 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22959046 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22959048 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22959085 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22959050 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22959007 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22959004 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22959010 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22959058 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22959066 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22959040 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22959018 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22959079 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22959006 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22959000 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22959081 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22959083 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22958989 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22959054 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22959023 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22959020 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22959045 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22958985 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22959017 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03258609 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22959065 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03315757 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03258613 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22959062 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22959006 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03364659 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22959055 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22959023 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22959057 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03823151 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22959056 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22959001 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22959070 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03258605 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03240925 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03258600 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03240926 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03722256 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03258602 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03284742 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22959025 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03392637 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22959013 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22959032 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03258599 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22959049 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22959004 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03284739 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22959019 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22959058 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03284740 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03250508 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22959027 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22959066 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03315753 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22959063 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03930566 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22959043 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03536913 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22959046 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03432583 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22959068 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22959011 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22959060 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22959067 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22959010 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03258606 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22959048 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22959007 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22959069 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03967531 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10513408 - -
          Affymetrix Mouse Gene 1.0 ST Array 10513411 - -
          Affymetrix Mouse Gene 1.0 ST Array 10513409 - -
          Affymetrix Mouse Gene 1.0 ST Array 10513410 - -
          Affymetrix HT MG-430 PM Array Plate 1456297_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1423476_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10513406 - -
          Mouse Genome 430 2.0 Array 1423476_PM_at - -
          Mouse Genome 430 2.0 Array 1423476_at - -
          Mouse Genome 430 2.0 Array 1456297_PM_at - -
          Mouse Genome 430 2.0 Array 1456297_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 30936_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 30936_at - -
          Affymetrix Mouse Clariom S Array PSR0400022428.mm.1 TC0400002765.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400022428.mm.1 NM_021053.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400022428.mm.1 NM_021053.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0400022429.mm.1 TC0400002765.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400022429.mm.1 NM_021053.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400022429.mm.1 NM_021053.4 Refseq
          Mouse Expression Set 430 1423476_PM_at - -
          Mouse Expression Set 430 1423476_at - -
          Mouse Expression Set 430 1456297_at - -
          Mouse Expression Set 430 1456297_PM_at - -
          Mouse Genome 430A 2.0 Array 1423476_PM_at - -
          Mouse Genome 430A 2.0 Array 1423476_at - -
          Affymetrix HT Mouse Genome 430A Array 1423476_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1423476_at - -
          Affymetrix HT Mouse Genome 430B Array 1456297_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1456297_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 5670309 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005670309 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2446648 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2446648 - -
          mouse-6 v1.1 (Illumina) scl24272.4_303-S - -
          Illumina Sentrix mouse-6.v1 bead array 5670309 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl24272.4_303-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5670309 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2446648 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2446648 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline