Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00498649_s1
          See other DNAJB7 GE Assays ›
          Gene Symbol:
          Dnajb7
          Gene Name:
          DnaJ (Hsp40) homolog, subfamily B, member 7
          Gene Aliases:
          4933424H20Rik, Dj5, mDj5
          Chromosome Location:
          Chr.15: 81407088 - 81408273 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00498649_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dnajb7
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_021317.2 NP_067292.2 1 360 102
          GenBank mRNA AB028855.1 - 1 360 102
          AK006201.1 - 1 386 102
          AK077186.1 - 1 389 102
          BC048738.1 - 1 349 102
          BC104396.1 - 1 307 102
          BC104397.1 - 1 307 102
          BC119478.2 - 1 307 102

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1500001921.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Dnajb7

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          chaperone binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P80423 BC048738.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P80423 BC104396.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P80423 BC104397.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P80423 BC119478.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P80423 AK077186.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P80423 BC048738.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P80423 BC104396.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P80423 BC104397.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P80423 BC119478.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P80423 AK077186.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P153453 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P80423 BC048738.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P80423 BC104396.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P80423 BC104397.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P80423 BC119478.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P80423 AK077186.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P80423 BC048738.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P80423 BC104396.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P80423 BC104397.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P80423 BC119478.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P80423 AK077186.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30377809 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30377786 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30377795 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30377814 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30377785 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30377805 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30377793 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30377789 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30377798 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30377815 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30377812 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30377802 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30377783 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30377780 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30377816 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30377781 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30377807 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30377810 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30377818 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30377792 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30377800 - -
          Whole Mouse Genome G4122A A_52_P80423 BC048738.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P80423 BC104396.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P80423 BC104397.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P80423 BC119478.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P80423 AK077186.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P80423 BC048738.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P80423 BC104396.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P80423 BC104397.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P80423 BC119478.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P80423 AK077186.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P129851 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12100784 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11903014 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30377812 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30377841 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30377725 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30377748 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30377810 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30377742 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30377800 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30377789 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30377720 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30377828 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30377839 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30377783 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30377730 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30377805 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30377824 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30377747 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30377746 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30377814 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30377819 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30377786 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30377737 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30377834 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30377781 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30377733 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30377798 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30377823 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30377778 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30377728 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30377740 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30377818 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30377780 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30377732 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30377830 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30377837 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30377726 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30377802 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30377833 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30377836 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30377735 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30377832 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30377815 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30377751 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30377736 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30377809 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30377757 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30377739 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30377828 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30377821 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11864284 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30377802 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30377807 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30377815 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30377786 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11864281 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30377816 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30377778 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11842874 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30377817 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30377792 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12334812 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30377814 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30377783 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30377781 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30377795 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30377798 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30377793 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30377826 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30377809 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30377800 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30377819 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30377805 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12182330 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30377789 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12458914 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30377830 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30377823 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10430741 - -
          Affymetrix Mouse Gene 1.0 ST Array 10430742 - -
          Affymetrix Mouse Gene 1.0 ST Array 10430740 - -
          Affymetrix HT MG-430 PM Array Plate 1450991_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10430739 - -
          Mouse Genome 430 2.0 Array 1450991_PM_at - -
          Mouse Genome 430 2.0 Array 1450991_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 57755_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 57755_at - -
          Affymetrix Mouse Clariom S Array PSR1500015101.mm.1 NM_021317.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500015101.mm.1 NM_021317.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1500015101.mm.1 TC1500001921.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500015102.mm.1 NM_021317.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500015102.mm.1 NM_021317.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1500015102.mm.1 TC1500001921.mm.1 MTA TC
          Mouse Expression Set 430 1450991_PM_at - -
          Mouse Expression Set 430 1450991_at - -
          Mouse Genome 430A 2.0 Array 1450991_PM_at - -
          Mouse Genome 430A 2.0 Array 1450991_at - -
          Affymetrix HT Mouse Genome 430A Array 1450991_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1450991_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 398330 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 1090593 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001090593 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2777208 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2777208 - -
          mouse-6 v1.1 (Illumina) scl46935.1.32_89-S - -
          Illumina Sentrix mouse-6.v1 bead array 1090593 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl46935.1.32_89-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1090593 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2777208 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2777208 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline