Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00498747_s1
          See other OLFR1264 GE Assays ›
          Gene Symbol:
          Olfr1264
          Gene Name:
          olfactory receptor 1264
          Gene Aliases:
          MOR236-1, Mor16, Ors16
          Chromosome Location:
          Chr.2: 90021138 - 90022064 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00498747_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr1264
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_021368.1 NP_067343.1 1 750 75

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr1264

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception
          detection of chemical stimulus involved in sensory perception of smell
          transmembrane signaling receptor activity
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2106544 NM_021368.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P105946 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2106544 NM_021368.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P473363 NM_021368.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P105946 - -
          Whole Mouse Genome G4122A A_51_P473363 NM_021368.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P133942 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P104571 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P105946 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415589 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415554 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415569 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415557 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415572 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415588 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415613 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415577 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415590 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415615 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415560 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415564 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415561 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415591 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415587 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415510 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415603 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415566 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415521 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415580 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415612 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415502 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415608 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415593 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415605 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415596 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415544 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415592 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415562 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415509 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415555 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415598 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415517 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415515 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415599 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415523 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415516 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415499 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415583 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415518 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415585 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415595 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415513 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415574 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415610 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415578 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415541 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21415539 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415607 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415593 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415585 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415557 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415603 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415555 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01649352 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415564 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415592 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415572 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415574 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415596 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415605 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01846489 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415561 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415583 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415577 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415612 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415615 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01986194 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415613 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415591 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415580 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415598 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415566 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415595 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415560 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415578 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415569 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415590 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02200538 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415588 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415554 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415610 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415562 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21415587 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01986199 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10484859 - -
          Affymetrix HT MG-430 PM Array Plate 1422385_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10484858 - -
          Mouse Genome 430 2.0 Array 1422385_PM_at - -
          Mouse Genome 430 2.0 Array 1422385_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 258206_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 258206_at - -
          Mouse Expression Set 430 1422385_PM_at - -
          Mouse Expression Set 430 1422385_at - -
          Murine Genome U74v2 Set 96985_PM_at - -
          Murine Genome U74v2 Set 96985_at - -
          Mouse Genome 430A 2.0 Array 1422385_PM_at - -
          Mouse Genome 430A 2.0 Array 1422385_at - -
          Affymetrix HT Mouse Genome 430A Array 1422385_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1422385_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 842372 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 1660025 - -
          Illumina mouse-6 v1.0 expression beadchip 1660025 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001660025 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2671035 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2671035 - -
          mouse-6 v1.1 (Illumina) scl4267.1.1_202-S - -
          Illumina Sentrix mouse-6.v1 bead array 1660025 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl4267.1.1_202-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1660025 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2671035 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2671035 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline