Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0500030433.mm.1 |
TC0500003376.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500030433.mm.1 |
NM_020044.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500030433.mm.1 |
NM_022964.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500030431.mm.1 |
TC0500003376.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500030431.mm.1 |
NM_020044.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500030431.mm.1 |
NM_022964.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500030434.mm.1 |
TC0500003376.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500030434.mm.1 |
NM_020044.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500030434.mm.1 |
NM_022964.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500030436.mm.1 |
TC0500003376.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500030436.mm.1 |
NM_020044.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500030436.mm.1 |
NM_022964.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500030435.mm.1 |
TC0500003376.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500030435.mm.1 |
NM_020044.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500030432.mm.1 |
TC0500003376.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0500030432.mm.1 |
NM_020044.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0500030432.mm.1 |
NM_022964.4 |
Refseq |