Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00499213_g1
          See other MRPS34 GE Assays ›
          Gene Symbol:
          Mrps34
          Gene Name:
          mitochondrial ribosomal protein S34
          Gene Aliases:
          0610007F04Rik, 5330430D13Rik, AV001970, Tce2
          Chromosome Location:
          Chr.17: 24895120 - 24896274 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00499213_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mrps34
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_023260.1 NP_075749.1 2 - 3 407 81
          GenBank mRNA AK002285.1 - 2 - 3 381 81
          AK004049.1 - 2 - 3 371 81
          AK007337.1 - 2 - 3 371 81
          AK216319.1 - 2 - 3 116 81
          BC024336.1 - 2 - 3 366 4985502 81

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700002770.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Mrps34

          Panther Classification:

          Molecular Function -

          ribosomal protein translational protein

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31100161 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31100159 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31100156 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31100158 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31100163 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31100164 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31100166 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31100165 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P443702 NM_023260.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P443702 AK004049.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P443702 AK007337.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P443702 AK019922.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P443702 BC024336.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P443702 NM_023260.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P443702 AK004049.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P443702 AK007337.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P443702 AK019922.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P443702 BC024336.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 664944 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P136831 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P141646 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P158729 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P443702 NM_023260.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P443702 AK004049.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P443702 AK007337.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P443702 AK019922.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P443702 BC024336.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P443702 NM_023260.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P443702 AK004049.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P443702 AK007337.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P443702 AK019922.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P443702 BC024336.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P443702 NM_023260.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P443702 AK004049.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P443702 AK007337.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P443702 AK019922.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P443702 BC024336.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P443702 NM_023260.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P443702 AK004049.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P443702 AK007337.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P443702 AK019922.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P443702 BC024336.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31100162 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31100138 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31100140 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31100132 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31100154 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31100141 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31100152 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31100137 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31100144 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31100157 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31100155 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31100143 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31100149 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31100161 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31100165 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31100159 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31100150 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31100166 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31100135 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31100147 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31100146 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31100167 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P141646 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P158729 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P136831 - -
          Whole Mouse Genome G4122A A_51_P443702 NM_023260.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P443702 AK004049.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P443702 AK007337.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P443702 AK019922.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P443702 BC024336.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P443702 NM_023260.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P443702 AK004049.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P443702 AK007337.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P443702 AK019922.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P443702 BC024336.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P109407 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P158729 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P136831 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P141646 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13230885 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13230868 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13230928 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13230923 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13230918 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13230886 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13230926 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31100164 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31100166 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31100156 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31100159 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31100158 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31100163 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31100161 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31100165 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100101 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100152 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100144 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100161 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100135 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100073 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100140 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100155 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100119 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100074 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100141 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100174 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100159 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100095 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100099 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100149 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100089 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100109 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100137 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100154 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100071 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100176 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100113 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100092 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100132 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100146 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100178 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100086 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100094 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100111 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100087 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100167 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100170 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100097 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100172 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100165 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100106 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100138 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100098 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100103 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100156 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100078 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31100165 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31100167 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13230907 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31100137 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13230928 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31100159 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13230861 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13230873 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31100155 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31100132 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13230871 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31100149 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31100166 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31100144 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31100163 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31100156 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31100141 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31100146 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31100157 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13230904 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10442642 - -
          Affymetrix Mouse Gene 1.0 ST Array 10442639 - -
          Affymetrix Mouse Gene 1.0 ST Array 10442640 - -
          Affymetrix Mouse Gene 1.0 ST Array 10442641 - -
          Affymetrix HT MG-430 PM Array Plate 1421971_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10442638 - -
          Mouse Genome 430 2.0 Array 1421971_PM_a_at - -
          Mouse Genome 430 2.0 Array 1421971_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 79044_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 79044_at - -
          Affymetrix Mouse Clariom S Array PSR1700002840.mm.1 NM_023260.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002840.mm.1 NM_023260.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002840.mm.1 TC1700002770.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700002841.mm.1 NM_023260.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002841.mm.1 NM_023260.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002841.mm.1 TC1700002770.mm.1 MTA TC
          Mouse Expression Set 430 1421971_a_at - -
          Mouse Expression Set 430 1421971_PM_a_at - -
          Murine Genome U74v2 Set 95730_at - -
          Murine Genome U74v2 Set 95730_PM_at - -
          Mouse Genome 430A 2.0 Array 1421971_a_at - -
          Mouse Genome 430A 2.0 Array 1421971_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1421971_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1421971_PM_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 905650 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 5700541 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005700541 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2655774 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2655774 - -
          mouse-6 v1.1 (Illumina) scl50998.1_28-S - -
          Illumina Sentrix mouse-6.v1 bead array 5700541 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl50998.1_28-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5700541 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2655774 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2655774 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl50998.1_28-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline