Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00499682_m1
          See other PPP1R11 GE Assays ›
          Gene Symbol:
          Ppp1r11
          Gene Name:
          protein phosphatase 1, regulatory (inhibitor) subunit 11
          Gene Aliases:
          1500041B02Rik, AV117883, Hcgv, Tcte5, Tctex-5, Tctex5
          Chromosome Location:
          Chr.17: 36948355 - 36951792 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00499682_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ppp1r11
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_029632.3 NP_083908.1 1 - 2 370 70
          GenBank mRNA AK005379.1 - 1 - 2 267 70
          AK077455.1 - 1 - 2 253 70
          AK089107.1 - 1 - 2 253 70
          AK170104.1 - 1 - 2 253 70
          BC027737.1 - 1 - 2 240 5003418 70
          EF528576.1 - 1 - 2 84 70

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700002041.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ppp1r11

          Panther Classification:

          Molecular Function -

          ubiquitin-protein ligase

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of phosphoprotein phosphatase activity
          negative regulation of protein dephosphorylation
          negative regulation of catalytic activity
          protein phosphatase inhibitor activity
          protein serine/threonine phosphatase inhibitor activity
          protein phosphatase 1 binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31166413 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31166375 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31166376 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31166412 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31166377 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31166408 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31166374 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31166378 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31166410 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31166379 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31166409 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P110791 AK162607.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P110791 AK170104.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P110791 BC027737.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P110791 NM_029632.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P110791 AK005379.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 947465 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 214902 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 755299 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 117078 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P114499 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P110791 AK162607.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P110791 AK170104.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P110791 BC027737.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P110791 NM_029632.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P110791 AK005379.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31166432 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31166435 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31166437 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31166453 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31166436 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31166447 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31166443 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31166448 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31166438 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31166450 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31166445 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31166456 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31166440 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31166452 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31166430 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31166457 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31166455 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31166442 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31166446 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31166433 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31166429 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P114499 - -
          Whole Mouse Genome G4122A A_51_P105320 AK162607.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P105320 BB842861.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P105320 NM_029632.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P105320 AK005379.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P105320 AK077455.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P110791 AK162607.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P110791 AK170104.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P110791 BC027737.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P110791 NM_029632.3 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P110791 AK005379.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P114499 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13447247 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13447239 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13447249 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13447233 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13447251 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31166412 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31166409 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31166379 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31166374 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31166410 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31166376 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31166375 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31166413 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31166408 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31166378 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31166377 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166360 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166371 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166377 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166373 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166426 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166480 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166457 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166406 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166358 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166470 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166366 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166443 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166452 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166424 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166473 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166379 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166369 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166421 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166395 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166488 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166446 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166493 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166482 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166456 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166370 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166393 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166430 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166469 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166428 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166490 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166381 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166362 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166435 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166390 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166387 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166433 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166404 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166383 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166467 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166453 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166496 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166436 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166391 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166475 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166494 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166402 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166485 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166384 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166440 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166364 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166375 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166471 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166500 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166478 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166442 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166502 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166450 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166385 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166397 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166437 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166432 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31166448 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13277703 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13447233 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31166443 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31166438 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31166453 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31166424 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31166456 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31166446 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31166437 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13277684 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31166426 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31166436 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31166450 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31166440 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31166442 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31166455 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31166435 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31166433 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31166448 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31166457 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31166432 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31166421 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31166423 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10450816 - -
          Affymetrix Mouse Gene 1.0 ST Array 10450817 - -
          Affymetrix Mouse Gene 1.0 ST Array 10450815 - -
          Affymetrix Mouse Gene 1.0 ST Array 10450818 - -
          Affymetrix HT MG-430 PM Array Plate 1417060_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1448565_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10450814 - -
          Mouse Genome 430 2.0 Array 1417060_PM_at - -
          Mouse Genome 430 2.0 Array 1417060_at - -
          Mouse Genome 430 2.0 Array 1448565_PM_at - -
          Mouse Genome 430 2.0 Array 1448565_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 76497_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 76497_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1700019455.mm.1 NM_029632.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019455.mm.1 NM_029632.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019455.mm.1 TC1700002041.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700019456.mm.1 NM_029632.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019456.mm.1 NM_029632.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019456.mm.1 TC1700002041.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700019449.mm.1 NM_029632.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019449.mm.1 NM_029632.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019449.mm.1 TC1700002041.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700019453.mm.1 NM_029632.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019453.mm.1 NM_029632.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019453.mm.1 TC1700002041.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700019454.mm.1 NM_029632.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019454.mm.1 NM_029632.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019454.mm.1 TC1700002041.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700019451.mm.1 NM_029632.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019451.mm.1 NM_029632.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019451.mm.1 TC1700002041.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700019452.mm.1 NM_029632.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019452.mm.1 NM_029632.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019452.mm.1 TC1700002041.mm.1 MTA TC
          Mouse Expression Set 430 1417060_at - -
          Mouse Expression Set 430 1417060_PM_at - -
          Mouse Expression Set 430 1448565_at - -
          Mouse Expression Set 430 1448565_PM_at - -
          Murine Genome U74v2 Set 166497_PM_at - -
          Murine Genome U74v2 Set 166497_at - -
          Mouse Genome 430A 2.0 Array 1417060_at - -
          Mouse Genome 430A 2.0 Array 1448565_PM_at - -
          Mouse Genome 430A 2.0 Array 1417060_PM_at - -
          Mouse Genome 430A 2.0 Array 1448565_at - -
          Affymetrix HT Mouse Genome 430A Array 1448565_at - -
          Affymetrix HT Mouse Genome 430A Array 1417060_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1417060_at - -
          Affymetrix HT Mouse Genome 430A Array 1448565_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 631849 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 3610072 - -
          MouseWG-6 v2 Expression BeadChip 100380113 - -
          Illumina mouse-6 v1.0 expression beadchip 3610072 - -
          Illumina mouse-6 v1.0 expression beadchip 100380113 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003610072 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2757489 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2757489 - -
          mouse-6 v1.1 (Illumina) scl49943.4_9-S - -
          mouse-6 v1.1 (Illumina) scl0076497.2_72-S - -
          Illumina Sentrix mouse-6.v1 bead array 3610072 - -
          Illumina Sentrix mouse-6.v1 bead array 100380113 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0076497.2_72-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 100380113 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3610072 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2757489 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2757489 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2816137 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0076497.2_72-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline