Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00499840_m1
          See other HIGD1B GE Assays ›
          Gene Symbol:
          Higd1b
          Gene Name:
          HIG1 domain family, member 1B
          Gene Aliases:
          2310056K19Rik
          Chromosome Location:
          Chr.11: 102835756 - 102844133 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00499840_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Higd1b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_080846.1 NP_543122.1 2 - 3 600 116
          GenBank mRNA AK009942.1 - 2 - 3 441 116
          BC005568.1 - 2 - 3 600 3967350 116

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100001700.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Higd1b

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2111523 NM_080846.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2111523 BC005568.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P112297 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2111523 NM_080846.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2111523 BC005568.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28081702 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28081721 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28081688 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28081716 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28081689 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28081686 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28081725 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28081722 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28081712 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28081684 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28081708 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28081698 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28081717 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28081703 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28081697 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28081692 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28081719 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28081729 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28081710 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28081715 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28081727 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28081691 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28081724 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28081705 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28081713 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28081693 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28081709 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28081704 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28081699 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P112297 - -
          Whole Mouse Genome G4122A A_52_P303343 NM_080846.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P303343 AK009942.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P303343 BC005568.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P110269 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P112297 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09146772 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09013592 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09013589 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09013604 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28081710 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28081703 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28081724 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28081688 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28081704 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28081697 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28081712 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28081680 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28081669 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28081709 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28081717 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28081691 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28081715 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28081702 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28081672 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28081682 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28081689 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28081729 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28081705 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28081722 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28081693 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28081708 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28081713 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28081719 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28081686 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28081727 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28081725 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28081696 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28081712 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28081701 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28081706 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28081705 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09168215 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28081699 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28081704 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09650809 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28081688 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28081713 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28081700 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28081719 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28081677 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09145484 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28081669 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09013603 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28081694 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28081715 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28081672 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09146772 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28081682 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28081686 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28081680 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28081674 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28081724 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28081717 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09168779 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28081675 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09712509 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28081684 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28081708 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28081689 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28081727 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28081691 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28081725 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28081722 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10381652 - -
          Affymetrix Mouse Gene 1.0 ST Array 10381651 - -
          Affymetrix Mouse Gene 1.0 ST Array 10381650 - -
          Affymetrix HT MG-430 PM Array Plate 1449860_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10381649 - -
          Mouse Genome 430 2.0 Array 1449860_PM_at - -
          Mouse Genome 430 2.0 Array 1449860_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 75689_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 75689_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1100016027.mm.1 NM_080846.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100016027.mm.1 NM_080846.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100016027.mm.1 TC1100001700.mm.1 MTA TC
          Mouse Expression Set 430 1449860_PM_at - -
          Mouse Expression Set 430 1449860_at - -
          Mouse Genome 430A 2.0 Array 1449860_at - -
          Mouse Genome 430A 2.0 Array 1449860_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1449860_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1449860_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 454877 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 2630088 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002630088 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1256099 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1256099 - -
          mouse-6 v1.1 (Illumina) scl40852.4.1_7-S - -
          Illumina Sentrix mouse-6.v1 bead array 2630088 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl40852.4.1_7-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2630088 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2852925 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2852925 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl40852.4.1_7-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline