Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00500199_m1
          See other ARF5 GE Assays ›
          Gene Symbol:
          Arf5
          Gene Name:
          ADP-ribosylation factor 5
          Gene Aliases:
          -
          Chromosome Location:
          Chr.6: 28423640 - 28426499 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00500199_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Arf5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_007480.1 NP_031506.1 4 - 5 384 59
          GenBank mRNA BC132452.1 - 5 - 6 372 40130729 59
          BC138756.1 - 5 - 6 372 8861778 59
          D87902.1 - 4 - 5 384 59

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600000213.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Arf5

          Panther Classification:

          Molecular Function -

          G-protein

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          small GTPase mediated signal transduction
          protein transport
          vesicle-mediated transport
          nucleotide binding
          protein binding
          GTP binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2067445 NM_007480.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2067445 D87902.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1976207 NM_007480.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1976207 BC132452.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1976207 BC138756.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1976207 D87902.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 204093 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2067445 NM_007480.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2067445 D87902.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1976207 NM_007480.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1976207 BC132452.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1976207 BC138756.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1976207 D87902.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P381801 NM_007480.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P381801 BC132452.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P381801 BC138756.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P381801 D87902.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24320127 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24320122 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24320111 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24320115 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24320125 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24320114 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24320120 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24320116 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24320109 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24320128 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24320107 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24320124 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24320112 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24320117 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24320108 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24320119 - -
          Whole Mouse Genome G4122A A_51_P381801 NM_007480.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P381801 BC132452.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P381801 BC138756.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P381801 D87902.1 mouse_transcriptome
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24320056 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24320139 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24320156 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24320114 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24320075 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24320128 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24320079 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24320161 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24320095 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24320068 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24320137 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24320048 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24320160 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24320147 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24320119 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24320170 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24320134 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24320117 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24320052 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24320131 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24320180 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24320063 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24320152 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24320125 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24320038 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24320149 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24320144 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24320162 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24320057 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24320099 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24320143 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24320177 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24320157 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24320151 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24320163 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24320182 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24320090 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24320120 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24320100 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24320097 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24320108 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24320127 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24320150 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24320058 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24320106 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24320092 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24320148 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24320124 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24320084 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24320089 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24320174 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24320107 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24320122 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24320066 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24320069 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24320154 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24320115 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24320166 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24320073 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24320158 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24320077 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24320087 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24320178 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24320061 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24320129 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24320045 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24320044 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24320112 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24320054 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24320082 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24320050 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24320141 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24320093 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24320104 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24320119 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05694157 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05694145 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24320112 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24320108 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05694148 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24320125 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24320103 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05058249 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24320106 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24320111 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24320116 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24320117 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24320114 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24320120 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10536750 - -
          Affymetrix Mouse Gene 1.0 ST Array 10536749 - -
          Affymetrix Mouse Gene 1.0 ST Array 10536751 - -
          Affymetrix Mouse Gene 1.0 ST Array 10536748 - -
          Affymetrix Mouse Gene 1.0 ST Array 10536747 - -
          Affymetrix Mouse Gene 1.1 ST Array 10536746 - -
          Mouse Genome 430 2.0 Array 1422807_PM_at - -
          Mouse Genome 430 2.0 Array 1422807_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 11844_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 11844_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0600001351.mm.1 TC0600000213.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600001351.mm.1 NM_007480.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600001351.mm.1 NM_007480.1 Refseq
          Mouse Expression Set 430 1422807_PM_at - -
          Mouse Expression Set 430 1422807_at - -
          Murine Genome U74v2 Set 92968_PM_at - -
          Murine Genome U74v2 Set 92968_at - -
          Mouse Genome 430A 2.0 Array 1422807_PM_at - -
          Mouse Genome 430A 2.0 Array 1422807_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 566025 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 4730369 - -
          Illumina mouse-6 v1.0 expression beadchip 4730369 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004730369 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2677712 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2677712 - -
          mouse-6 v1.1 (Illumina) scl011844.1_12-S - -
          Illumina Sentrix mouse-6.v1 bead array 4730369 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl011844.1_12-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4730369 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2677712 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2677712 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl011844.1_12-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline