Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Gene Symbol: Ocln | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008756.2 | NP_032782.1 | 5 - 6 | 1219 | 83 | |
GenBank mRNA | AK019880.1 | - | 5 - 6 | 1171 | 83 | |
AK033532.1 | - | 5 - 6 | 1152 | 83 | ||
AK042576.1 | - | 5 - 6 | 1219 | 83 | ||
AK145798.1 | - | 3 - 4 | 378 | 83 | ||
BC138679.1 | - | 5 - 6 | 1202 | 8861700 | 83 | |
BC138680.1 | - | 5 - 6 | 1202 | 8861701 | 83 | |
BC145371.1 | - | 5 - 6 | 1217 | 9053929 | 83 | |
U49185.1 | - | 5 - 6 | 1262 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons |