Search
Search

Pcsk2| Gene Symbol: Pcsk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008792.4 | NP_032818.1 | 1 - 2 | 534 | 75 | |
| GenBank mRNA | AF483512.1 | - | 1 - 2 | 176 | 75 | |
| AF483513.1 | - | 1 - 2 | 176 | 75 | ||
| AK039783.1 | - | 1 - 2 | 397 | 75 | ||
| AK133620.1 | - | 1 - 2 | 535 | 75 | ||
| AK158493.1 | - | 1 - 2 | 482 | 75 | ||
| AK158598.1 | - | 1 - 2 | 596 | 75 | ||
| AK161585.1 | - | 1 - 2 | 475 | 75 | ||
| BC052013.1 | - | 1 - 2 | 451 | 5707923 | 75 | |
| BC057348.1 | - | 1 - 2 | 489 | 6827695 | 75 | |
| M55669.1 | - | 1 - 2 | 260 | 75 | ||
Set Membership: |
|