Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00501325_m1
          See other RCVRN GE Assays ›
          Gene Symbol:
          Rcvrn
          Gene Name:
          recoverin
          Gene Aliases:
          CAR, S-modulin
          Chromosome Location:
          Chr.11: 67695326 - 67703355 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00501325_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rcvrn
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_009038.2 NP_033064.1 1 - 2 473 80
          GenBank mRNA BC110564.2 - 1 - 2 466 40050126 80
          CK617354.1 - 1 - 2 433 80
          X66196.1 - 1 - 2 481 80

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100000861.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rcvrn

          Gene Ontology Categories:

          Function(s) Process(es)

          visual perception
          phototransduction
          response to stimulus
          regulation of calcium ion transport
          calcium ion binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27884873 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27884875 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27884874 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P102910 NM_009038.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P102910 CK617615.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P102910 CK627353.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P102910 X66196.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 906094 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 13552 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 400502 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 72011 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 71406 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P133219 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P102910 NM_009038.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P102910 CK617615.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P102910 CK627353.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P102910 X66196.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P503922 X66196.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27884875 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27884865 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27884858 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27884857 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27884845 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27884861 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27884881 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27884871 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27884853 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27884863 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27884859 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27884877 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27884868 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27884880 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27884873 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27884886 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27884872 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27884885 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27884882 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27884864 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27884850 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27884855 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27884884 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27884847 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27884867 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27884870 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27884889 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02352254 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P133219 - -
          Whole Mouse Genome G4122A A_51_P503922 X66196.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P102910 NM_009038.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P102910 CK617615.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P102910 CK627353.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P102910 X66196.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P133219 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09742944 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09742940 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09742936 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09742946 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27884874 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27884873 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27884875 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27884919 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27884875 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27884938 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27884842 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27884902 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27884856 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27884915 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27884870 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27884889 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27884935 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27884896 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27884905 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27884913 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27884864 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27884899 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27884886 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27884937 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27884859 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27884882 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27884871 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27884891 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27884921 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27884884 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27884857 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27884931 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27884845 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27884908 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27884873 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27884926 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27884877 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27884897 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27884933 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27884843 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27884923 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27884911 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27884927 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27884904 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27884847 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27884868 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27884880 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27884850 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27884881 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27884917 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27884853 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27884895 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27884839 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27884924 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27884900 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27884907 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27884863 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27884850 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27884911 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27884897 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09537835 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27884901 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27884845 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09063948 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27884921 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27884910 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27884856 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27884913 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27884917 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27884905 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27884877 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27884908 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27884868 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27884881 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27884847 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27884895 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27884843 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27884853 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27884919 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27884904 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27884889 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27884861 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27884864 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27884899 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09586802 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27884865 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09112682 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27884909 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27884866 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27884857 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27884880 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27884855 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27884882 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27884842 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09586798 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27884902 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27884863 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27884884 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27884918 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09086177 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09184765 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27884891 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27884873 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27884859 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09742946 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27884875 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27884915 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09103191 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27884896 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27884870 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10377236 - -
          Affymetrix Mouse Gene 1.0 ST Array 10377238 - -
          Affymetrix Mouse Gene 1.0 ST Array 10377237 - -
          Affymetrix Mouse Gene 1.0 ST Array 10377239 - -
          Affymetrix HT MG-430 PM Array Plate 1450215_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10377235 - -
          Mouse Genome 430 2.0 Array 1450215_at - -
          Mouse Genome 430 2.0 Array 1450215_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 19674_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 19674_at - -
          Affymetrix Mouse Clariom S Array PSR1100007140.mm.1 NM_009038.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100007140.mm.1 NM_009038.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100007140.mm.1 TC1100000861.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100007139.mm.1 NM_009038.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100007139.mm.1 NM_009038.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100007139.mm.1 TC1100000861.mm.1 MTA TC
          Mouse Expression Set 430 1450215_PM_at - -
          Mouse Expression Set 430 1450215_at - -
          Murine Genome U74v2 Set 101151_PM_at - -
          Murine Genome U74v2 Set 101151_at - -
          Mouse Genome 430A 2.0 Array 1450215_at - -
          Mouse Genome 430A 2.0 Array 1450215_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1450215_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1450215_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 631184 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 4010279 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004010279 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2661924 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2661924 - -
          mouse-6 v1.1 (Illumina) scl41402.3.1_0-S - -
          Illumina Sentrix mouse-6.v1 bead array 4010279 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl41402.3.1_0-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4010279 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2661924 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2661924 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl41402.3.1_0-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline