Search
Search
| Gene Symbol: Rfx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009055.4 | NP_033081.3 | 2 - 3 | 553 | 75 | |
| GenBank mRNA | AK036208.1 | - | 2 - 3 | 555 | 75 | |
| AK044607.1 | - | 2 - 3 | 554 | 75 | ||
| AK147790.1 | - | 2 - 3 | 556 | 75 | ||
| BB642620.1 | - | 2 - 3 | 554 | 75 | ||
| BC057018.1 | - | 2 - 3 | 532 | 5705637 | 75 | |
| X76088.1 | - | 2 - 3 | 460 | 75 | ||
Set Membership: |
|