Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00501400_m1
          See other RIT1 GE Assays ›
          Gene Symbol:
          Rit1
          Gene Name:
          Ras-like without CAAX 1
          Gene Aliases:
          RIBB, ROC1, Rit
          Chromosome Location:
          Chr.3: 88716840 - 88731048 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00501400_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rit1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001163310.1 NP_001156782.1 2 - 3 160 83
          NM_009069.4 NP_033095.1 3 - 4 305 83
          NR_028056.1 3 - 4 334 83
          GenBank mRNA AK005357.1 - 3 - 4 266 83
          AK014126.1 - 3 - 4 296 83
          AK018785.1 - 3 - 4 253 83
          AK164196.1 - 3 - 4 296 83
          AK168254.1 - 3 - 4 264 83
          BC012694.1 - 3 - 4 265 4023732 83
          BY284832.1 - 3 - 4 308 83
          CV562852.1 - 3 - 4 273 83
          CX242151.1 - 2 - 3 108 83
          U71205.1 - 3 - 4 242 83

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0300000736.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rit1

          Panther Classification:

          Molecular Function -

          small GTPase

          Gene Ontology Categories:

          Function(s) Process(es)

          signal transduction
          small GTPase mediated signal transduction
          Ras protein signal transduction
          nucleotide binding
          protein binding
          GTP binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22315261 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22315262 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22315263 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22315264 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P236175 BC012694.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P236175 U71205.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P236175 AK018785.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P236175 AK164196.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P236175 AK168254.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P236175 BC012694.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P236175 U71205.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P236175 AK018785.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P236175 AK164196.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P236175 AK168254.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 388186 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 355008 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 427491 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 754283 AK154420.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 754283 BC061154.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 754283 D87757.2 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 754283 M94584.2 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 754283 NM_009892.1 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 754283 U56900.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 754283 XM_884904.3 refseq_mrna_mouse_XM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 798014 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 961490 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 890666 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 732334 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 76561 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 136739 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P171908 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P163475 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P236175 BC012694.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P236175 U71205.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P236175 AK018785.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P236175 AK164196.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P236175 AK168254.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P236175 BC012694.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P236175 U71205.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P236175 AK018785.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P236175 AK164196.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P236175 AK168254.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P236175 BC012694.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P236175 U71205.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P236175 AK018785.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P236175 AK164196.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P236175 AK168254.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P236175 BC012694.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P236175 U71205.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P236175 AK018785.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P236175 AK164196.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P236175 AK168254.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22315276 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22315274 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22315265 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22315270 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22315280 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22315254 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22315267 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22315259 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22315262 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22315252 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22315272 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22315264 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22315261 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22315278 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22315268 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22315257 - -
          Whole Mouse Genome G4122A A_51_P236175 BC012694.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P236175 U71205.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P236175 AK018785.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P236175 AK164196.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P236175 AK168254.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P236175 BC012694.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P236175 U71205.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P236175 AK018785.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P236175 AK164196.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P236175 AK168254.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P515370 AK018785.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P515370 AK168254.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P515370 NR_028056.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P515370 NM_001163310.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P515370 NM_009069.4 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P117944 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02490481 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22315264 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22315263 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22315262 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22315261 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22315307 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22315325 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22315357 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22315280 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22315288 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22315274 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22315313 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22315276 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22315312 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22315353 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22315282 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22315332 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22315327 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22315334 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22315264 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22315361 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22315359 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22315363 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22315301 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22315262 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22315339 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22315278 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22315281 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22315308 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22315298 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22315272 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22315319 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22315277 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22315323 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22315286 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22315275 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22315366 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22315347 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22315331 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22315257 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22315367 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22315293 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22315265 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22315310 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22315342 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22315355 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22315309 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22315270 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22315267 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22315344 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22315283 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22315337 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02695883 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22315276 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02695879 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22315348 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22315295 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22315251 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22315264 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22315284 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22315281 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22315286 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22315334 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22315267 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22315291 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22315254 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22315278 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22315339 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22315293 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22315297 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22315296 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22315301 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22315298 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22315270 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22315262 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22315259 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22315302 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02564956 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22315275 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22315342 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22315272 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22315344 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22315257 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02695886 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02819954 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22315255 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22315333 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22315265 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22315274 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22315288 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02564959 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02624730 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10493312 - -
          Affymetrix Mouse Gene 1.0 ST Array 10493313 - -
          Affymetrix Mouse Gene 1.0 ST Array 10493315 - -
          Affymetrix Mouse Gene 1.0 ST Array 10493314 - -
          Affymetrix Mouse Gene 1.0 ST Array 10493311 - -
          Affymetrix Mouse Gene 1.0 ST Array 10493310 - -
          Affymetrix HT MG-430 PM Array Plate 1420540_PM_a_at - -
          Affymetrix HT MG-430 PM Array Plate 1428710_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10493309 - -
          Mouse Genome 430 2.0 Array 1428710_PM_at - -
          Mouse Genome 430 2.0 Array 1420540_PM_a_at - -
          Mouse Genome 430 2.0 Array 1428710_at - -
          Mouse Genome 430 2.0 Array 1420540_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 19769_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 19769_at - -
          Affymetrix Mouse Clariom S Array PSR0300005324.mm.1 NM_001163310.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300005324.mm.1 NM_001163310.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300005324.mm.1 NM_009069.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300005324.mm.1 NM_009069.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0300005324.mm.1 NR_028056.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300005324.mm.1 NR_028056.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300005324.mm.1 TC0300000736.mm.1 MTA TC
          Mouse Expression Set 430 1428710_at - -
          Mouse Expression Set 430 1420540_PM_a_at - -
          Mouse Expression Set 430 1420540_a_at - -
          Mouse Expression Set 430 1428710_PM_at - -
          Murine Genome U74v2 Set 92376_at - -
          Murine Genome U74v2 Set 92376_PM_at - -
          Mouse Genome 430A 2.0 Array 1420540_a_at - -
          Mouse Genome 430A 2.0 Array 1420540_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1428710_PM_at - -
          Mouse Genome 430A 2.0 Array 1428710_at - -
          Affymetrix HT Mouse Genome 430A Array 1420540_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1420540_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1428710_at - -
          Affymetrix HT Mouse Genome 430A Array 1428710_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 568190 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 102480010 - -
          MouseWG-6 v2 Expression BeadChip 5390338 - -
          Illumina mouse-6 v1.0 expression beadchip 5390338 - -
          Illumina mouse-6 v1.0 expression beadchip 102480010 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005390338 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1228359 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1228359 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2489666 - -
          mouse-6 v1.1 (Illumina) scl22992.4_72-S - -
          mouse-6 v1.1 (Illumina) scl0019769.2_219-S - -
          Illumina Sentrix mouse-6.v1 bead array 5390338 - -
          Illumina Sentrix mouse-6.v1 bead array 102480010 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0019769.2_219-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 102480010 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5390338 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1228359 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1228359 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0019769.2_219-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline