Search
Search
| Gene Symbol: Rps5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009095.2 | NP_033121.2 | 2 - 3 | 188 | 69 | |
| GenBank mRNA | AK002235.1 | - | 2 - 3 | 182 | 69 | |
| AK009663.1 | - | 2 - 3 | 182 | 69 | ||
| AK010681.1 | - | 2 - 3 | 182 | 69 | ||
| AK012424.1 | - | 2 - 3 | 182 | 69 | ||
| AK012481.1 | - | 2 - 3 | 182 | 69 | ||
| AK020477.1 | - | 2 - 3 | 181 | 69 | ||
| AK020754.1 | - | 2 - 3 | 182 | 69 | ||
| AK050611.1 | - | 2 - 3 | 182 | 69 | ||
| AK050622.1 | - | 2 - 3 | 182 | 69 | ||
| AK152449.1 | - | 2 - 3 | 182 | 69 | ||
| AK160552.1 | - | 2 - 3 | 182 | 69 | ||
| AK164079.1 | - | 2 - 3 | 182 | 69 | ||
| AK167533.1 | - | 2 - 3 | 182 | 69 | ||
| AK168044.1 | - | 2 - 3 | 182 | 69 | ||
| AK168106.1 | - | 2 - 3 | 182 | 69 | ||
| AK169016.1 | - | 2 - 3 | 182 | 69 | ||
| AK172446.1 | - | 2 - 3 | 182 | 69 | ||
| BC020401.1 | - | 2 - 3 | 162 | 69 | ||
| BC058690.1 | - | 2 - 3 | 169 | 6814814 | 69 | |
| U78085.1 | - | 2 - 3 | 174 | 69 | ||
| Y12431.1 | - | 2 - 3 | 164 | 69 | ||
Set Membership: |
|