Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00501568_m1
          See other CAPNS1 GE Assays ›
          Gene Symbol:
          Capns1
          Gene Name:
          calpain, small subunit 1
          Gene Aliases:
          Capa-4, Capa4, Capn4, D7Ertd146e
          Chromosome Location:
          Chr.7: 30186942 - 30198794 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00501568_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Capns1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_009795.3 NP_033925.2 6 - 7 521 116
          GenBank mRNA AF058298.1 - 5 - 6 500 116
          AK028092.1 - 4 - 5 267 116
          AK132819.1 - 5 - 6 331 116
          BC018352.1 - 6 - 7 515 3156021 116
          BC090988.1 - 6 - 7 521 30246016 116

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700002745.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Protease Mammalian Gene Collection Recommended Assay Mus musculus Mouse Alzheimer's Disease 1700 Inventoried Mouse_Alzheimer Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Capns1

          Gene Ontology Categories:

          Function(s) Process(es)

          proteolysis
          calcium-dependent cysteine-type endopeptidase activity
          calcium ion binding
          metal ion binding
          protein heterodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2026400 AK132819.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2026400 BC018352.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2026400 BC090988.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2026400 NM_009795.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2026400 AF058298.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2124528 BC090988.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2124528 NM_009795.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2124528 AK028092.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 154911 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 615245 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 436079 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 346710 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 197102 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 143913 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P152464 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2124528 BC090988.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2124528 NM_009795.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2124528 AK028092.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2026400 AK132819.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2026400 BC018352.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2026400 BC090988.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2026400 NM_009795.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2026400 AF058298.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25030305 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25030334 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25030301 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25030308 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25030326 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25030293 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25030315 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25030330 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25030340 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25030345 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25030317 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25030300 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25030321 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25030327 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25030295 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25030337 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25030333 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25030335 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25030302 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25030319 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25030303 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25030342 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25030332 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25030341 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25030311 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25030328 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25030309 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25030297 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25030344 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25030294 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25030323 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25030313 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25030316 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25030298 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25030347 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25030304 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P152464 - -
          Whole Mouse Genome G4122A A_51_P399538 AF058298.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P125368 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P152464 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06404062 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06478761 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05893357 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05970380 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06018142 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030283 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030351 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030355 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030334 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030319 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030297 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030310 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030347 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030253 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030317 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030304 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030302 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030313 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030305 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030300 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030309 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030324 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030298 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030252 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030327 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030308 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030250 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030361 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030262 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030341 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030284 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030275 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030330 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030354 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030279 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030292 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030264 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030259 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030323 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030326 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030344 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030303 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030267 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030333 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030249 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030295 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030348 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030345 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030358 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030288 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030352 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030280 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030316 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030364 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030261 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030290 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030289 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030336 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030315 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030263 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030332 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030356 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030266 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030294 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030256 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030254 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030271 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25030304 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06037838 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25030295 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25030303 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25030285 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25030289 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25030323 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06061007 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06193031 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25030284 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06230676 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05990103 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25030279 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25030328 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25030283 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06663268 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05953952 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25030300 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06549988 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06363085 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25030337 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25030340 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25030282 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25030290 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25030354 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06061009 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25030330 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25030326 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25030294 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06017755 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25030329 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25030315 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25030336 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05929506 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06549986 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25030302 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06478761 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25030275 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06253060 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06402327 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25030319 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25030288 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06037841 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05990113 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06404062 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05990111 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25030347 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05970380 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25030292 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25030316 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06102662 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25030350 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25030297 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25030313 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05990105 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06037836 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10561850 - -
          Affymetrix Mouse Gene 1.0 ST Array 10561849 - -
          Affymetrix Mouse Gene 1.0 ST Array 10561843 - -
          Affymetrix Mouse Gene 1.0 ST Array 10561853 - -
          Affymetrix Mouse Gene 1.0 ST Array 10561845 - -
          Affymetrix Mouse Gene 1.0 ST Array 10561852 - -
          Affymetrix Mouse Gene 1.0 ST Array 10561846 - -
          Affymetrix Mouse Gene 1.0 ST Array 10561848 - -
          Affymetrix Mouse Gene 1.0 ST Array 10561851 - -
          Affymetrix Mouse Gene 1.0 ST Array 10561847 - -
          Affymetrix HT MG-430 PM Array Plate 1426400_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10561842 - -
          Mouse Genome 430 2.0 Array 1426400_a_at - -
          Mouse Genome 430 2.0 Array 1426400_PM_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 12336_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 12336_at - -
          Affymetrix Mouse Clariom S Array PSR0700024324.mm.1 TC0700002745.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700024324.mm.1 NM_009795.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024329.mm.1 TC0700002745.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700024329.mm.1 NM_009795.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024330.mm.1 TC0700002745.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700024330.mm.1 NM_009795.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024320.mm.1 TC0700002745.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700024320.mm.1 NM_009795.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024322.mm.1 TC0700002745.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700024322.mm.1 NM_009795.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024321.mm.1 TC0700002745.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700024321.mm.1 NM_009795.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024325.mm.1 TC0700002745.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700024325.mm.1 NM_009795.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024327.mm.1 TC0700002745.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700024327.mm.1 NM_009795.4 Refseq
          Mouse Expression Set 430 1426400_a_at - -
          Mouse Expression Set 430 1426400_PM_a_at - -
          Murine Genome U74v2 Set 100610_PM_at - -
          Murine Genome U74v2 Set 100610_at - -
          Mouse Genome 430A 2.0 Array 1426400_a_at - -
          Mouse Genome 430A 2.0 Array 1426400_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1426400_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1426400_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 435008 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 3940113 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003940113 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2772249 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2772249 - -
          mouse-6 v1.1 (Illumina) scl31535.10.6_2-S - -
          Illumina Sentrix mouse-6.v1 bead array 3940113 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl31535.10.6_2-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3940113 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2772248 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2772248 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl31535.10.6_2-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline