Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00501637_s1
          See other CYP8B1 GE Assays ›
          Gene Symbol:
          Cyp8b1
          Gene Name:
          cytochrome P450, family 8, subfamily b, polypeptide 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.9: 121914356 - 121916305 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00501637_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information
          • Citations (1)

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cyp8b1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_010012.3 NP_034142.3 1 294 69
          GenBank mRNA AF090317.1 - 1 280 69
          BC010973.1 - 1 304 4161038 69
          BC010974.1 - 1 297 4162964 69
          BC049969.1 - 1 279 4158774 69

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0900003276.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection P450 Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cyp8b1

          Gene Ontology Categories:

          Function(s) Process(es)

          lipid metabolic process
          oxidation-reduction process
          monooxygenase activity
          iron ion binding
          sterol 12-alpha-hydroxylase activity
          oxidoreductase activity
          oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen
          heme binding
          7alpha-hydroxycholest-4-en-3-one 12alpha-hydroxylase activity
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P266618 BC010974.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P266618 BC049969.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P266618 NM_010012.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P266618 AF090317.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P266618 BC010973.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P266618 BC010974.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P266618 BC049969.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P266618 NM_010012.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P266618 AF090317.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P266618 BC010973.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 841255 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P161987 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P266618 BC010974.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P266618 BC049969.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P266618 NM_010012.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P266618 AF090317.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P266618 BC010973.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P266618 BC010974.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P266618 BC049969.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P266618 NM_010012.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P266618 AF090317.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P266618 BC010973.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P266618 BC010974.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P266618 BC049969.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P266618 NM_010012.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P266618 AF090317.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P266618 BC010973.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P266618 BC010974.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P266618 BC049969.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P266618 NM_010012.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P266618 AF090317.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P266618 BC010973.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26886364 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26886357 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26886344 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26886356 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26886348 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26886350 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26886366 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26886374 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26886347 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26886355 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26886369 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26886349 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26886360 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26886342 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26886354 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26886371 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26886337 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26886372 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26886339 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26886361 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26886362 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26886358 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26886334 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26886352 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P161987 - -
          Whole Mouse Genome G4122A A_51_P266618 BC010974.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P266618 BC049969.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P266618 NM_010012.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P266618 AF090317.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P266618 BC010973.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P266618 BC010974.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P266618 BC049969.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P266618 NM_010012.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P266618 AF090317.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P266618 BC010973.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P112640 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P161987 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08173107 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08173079 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08173099 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26886277 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26886344 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26886331 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26886281 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26886357 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26886325 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26886337 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26886358 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26886380 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26886286 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26886334 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26886322 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26886387 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26886330 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26886339 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26886350 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26886392 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26886347 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26886366 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26886364 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26886282 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26886352 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26886394 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26886273 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26886362 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26886329 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26886283 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26886382 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26886342 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26886276 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26886360 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26886361 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26886372 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26886355 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26886349 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26886274 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26886369 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26886380 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26886366 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08019210 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26886349 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26886360 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07627285 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26886358 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26886352 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26886330 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26886355 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26886347 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26886372 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26886337 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26886377 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26886331 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26886364 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07581426 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26886362 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26886374 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26886329 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08173098 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26886381 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26886378 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08173094 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26886342 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07570109 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26886357 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26886339 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26886325 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08173073 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26886334 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26886350 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10597876 - -
          Affymetrix Mouse Gene 1.0 ST Array 10597877 - -
          Affymetrix HT MG-430 PM Array Plate 1449309_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10597875 - -
          Mouse Genome 430 2.0 Array 1449309_at - -
          Mouse Genome 430 2.0 Array 1449309_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 13124_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 13124_at - -
          Affymetrix Mouse Clariom S Array PSR0900027646.mm.1 NM_010012.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900027646.mm.1 NM_010012.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0900027646.mm.1 TC0900003276.mm.1 MTA TC
          Mouse Expression Set 430 1449309_at - -
          Mouse Expression Set 430 1449309_PM_at - -
          Murine Genome U74v2 Set 103284_PM_at - -
          Murine Genome U74v2 Set 103284_at - -
          Mouse Genome 430A 2.0 Array 1449309_PM_at - -
          Mouse Genome 430A 2.0 Array 1449309_at - -
          Affymetrix HT Mouse Genome 430A Array 1449309_at - -
          Affymetrix HT Mouse Genome 430A Array 1449309_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 517553 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2350369 - -
          Illumina mouse-6 v1.0 expression beadchip 2350369 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002350369 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2627744 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2627744 - -
          mouse-6 v1.1 (Illumina) scl35191.1.7_235-S - -
          Illumina Sentrix mouse-6.v1 bead array 2350369 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl35191.1.7_235-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2350369 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2627744 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2627744 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl35191.1.7_235-S - -

          Back To Top
           

          Citations (1)

          Fibroblast growth factor 21 corrects obesity in mice.
          Authors
          Coskun T, Bina HA, Schneider MA, Dunbar JD, Hu CC, Chen Y, Moller DE, Kharitonenkov A
          Journal
          Endocrinology - Page No 6018 (Mon Dec 01 00:00:00 EST 2008)
          Volume (Year)
          149 (2008)
          Product Usage
          -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline