Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00501996_m1
          See other IP6K1 GE Assays ›
          Gene Symbol:
          Ip6k1
          Gene Name:
          inositol hexaphosphate kinase 1
          Gene Aliases:
          1200016D08Rik, AU022776, Ihpk1, InsP6, InsP6k1, Itpk6
          Chromosome Location:
          Chr.9: 108002648 - 108048782 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00501996_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ip6k1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_013785.2 NP_038813.2 3 - 4 736 68
          GenBank mRNA AF177144.1 - 2 - 3 436 68
          AK088872.1 - 3 - 4 753 68
          AK171310.1 - 3 - 4 741 68
          AK172921.1 - 3 - 4 795 68
          AK193897.1 - 1 - 2 80 68
          BC053030.1 - 3 - 4 741 68
          BC059006.1 - 3 - 4 736 6827203 68

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0900003337.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ip6k1

          Panther Classification:

          Molecular Function -

          kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          inositol metabolic process
          phosphorylation
          inositol phosphate biosynthetic process
          phosphatidylinositol phosphorylation
          nucleotide binding
          inositol hexakisphosphate 5-kinase activity
          ATP binding
          inositol-1,4,5-trisphosphate 3-kinase activity
          kinase activity
          transferase activity
          inositol hexakisphosphate 1-kinase activity
          inositol hexakisphosphate 3-kinase activity

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0900011125.mm.1 NM_013785.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900011125.mm.1 NM_013785.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0900011125.mm.1 TC0900003337.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900011107.mm.1 NM_013785.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900011107.mm.1 NM_013785.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0900011107.mm.1 TC0900003337.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900011108.mm.1 NM_013785.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900011108.mm.1 NM_013785.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0900011108.mm.1 TC0900003337.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900011116.mm.1 NM_013785.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900011116.mm.1 NM_013785.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0900011116.mm.1 TC0900003337.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900011129.mm.1 NM_013785.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900011129.mm.1 NM_013785.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0900011129.mm.1 TC0900003337.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900011115.mm.1 NM_013785.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900011115.mm.1 NM_013785.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0900011115.mm.1 TC0900003337.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900011124.mm.1 NM_013785.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900011124.mm.1 NM_013785.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0900011124.mm.1 TC0900003337.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900028049.mm.1 NM_013785.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900028049.mm.1 NM_013785.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0900028049.mm.1 TC0900003337.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900011126.mm.1 NM_013785.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900011126.mm.1 NM_013785.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0900011126.mm.1 TC0900003337.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900011127.mm.1 NM_013785.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900011127.mm.1 NM_013785.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0900011127.mm.1 TC0900003337.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900011118.mm.1 NM_013785.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900011118.mm.1 NM_013785.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0900011118.mm.1 TC0900003337.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900011128.mm.1 NM_013785.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900011128.mm.1 NM_013785.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0900011128.mm.1 TC0900003337.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 911780 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-jwmpg:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0