Search
Search

Tmem131| Gene Symbol: Tmem131 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018872.2 | NP_061360.2 | 36 - 37 | 5059 | 79 | |
| GenBank mRNA | AB093221.1 | - | 32 - 33 | 4495 | 79 | |
| AF060565.1 | - | 36 - 37 | 4761 | 79 | ||
| BC120906.2 | - | 6 - 7 | 1018 | 79 | ||
| BC139734.1 | - | 6 - 7 | 1032 | 79 | ||
Set Membership: |
|