Search
Search

Pidd1| Gene Symbol: Pidd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022654.1 | NP_073145.1 | 1 - 2 | 387 | 104 | |
| GenBank mRNA | AF274973.1 | - | 1 - 2 | 387 | 104 | |
| AK004702.1 | - | 2 - 3 | 425 | 104 | ||
| AK087880.1 | - | 426 | 103 | |||
| AK156863.1 | - | 3 - 4 | 375 | 104 | ||
| BC032202.1 | - | 1 - 2 | 406 | 104 | ||
| BC056987.1 | - | 2 - 3 | 409 | 104 | ||
| BC089373.1 | - | 1 - 2 | 396 | 104 | ||
| BC145314.1 | - | 1 - 2 | 352 | 104 | ||
| BC145315.1 | - | 1 - 2 | 352 | 104 | ||
| BC145857.1 | - | 1 - 2 | 352 | 40131160 | 104 | |
Set Membership: |
|