Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00502719_m1
          See other RSRP1 GE Assays ›
          Gene Symbol:
          Rsrp1
          Gene Name:
          arginine/serine rich protein 1
          Gene Aliases:
          2700043I21Rik, D4Ucla2, D4Wsu53e
          Chromosome Location:
          Chr.4: 134923625 - 134927370 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00502719_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rsrp1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_023665.3 NP_076154.3 2 - 3 747 73
          GenBank mRNA AB041555.1 - 2 - 3 727 73
          AK003799.1 - 2 - 3 748 73
          AK012363.1 - 2 - 3 748 73
          AK028145.1 - 2 - 3 749 73
          AK029529.1 - 2 - 3 747 73
          AK150024.1 - 2 - 3 748 73
          AK150665.1 - 2 - 3 727 73
          AK152736.1 - 2 - 3 727 73
          AK160993.1 - 2 - 3 748 73
          AK169598.1 - 2 - 3 749 73
          AK169760.1 - 2 - 3 748 73
          AK178549.1 - 1 - 2 93 73
          AK206224.1 - 1 - 2 93 73
          BC043057.1 - 2 - 3 732 6419597 73
          BC046231.1 - 2 - 3 649 6813388 73
          BC054728.1 - 2 - 3 720 6419723 73
          BC056986.1 - 2 - 3 681 6816649 73

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400001646.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rsrp1

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23325562 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23325561 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23325563 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23325564 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23325560 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2131766 BC054728.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2131766 BC056986.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2131766 BC046231.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2131766 BC043057.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2131766 AK169760.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 836611 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 72611 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2131766 BC054728.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2131766 BC056986.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2131766 BC046231.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2131766 BC043057.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2131766 AK169760.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23325581 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23325561 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23325559 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23325565 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23325570 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23325564 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23325582 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23325560 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23325573 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23325571 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23325587 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23325586 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23325575 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23325567 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23325583 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23325576 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23325569 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23325574 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23325566 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23325577 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23325563 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23325579 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00995655 - -
          Whole Mouse Genome G4122A A_51_P346715 BC054728.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P346715 BC056986.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P346715 BC046231.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P346715 BC043057.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P346715 AK169760.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P113539 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P00995655 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03778611 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23325563 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23325561 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23325562 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23325560 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23325564 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325515 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325542 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325638 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325539 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325525 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325581 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325617 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325641 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325604 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325531 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325613 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325575 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325618 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325624 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325563 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325523 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325533 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325608 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325564 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325599 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325582 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325591 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325577 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325574 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325628 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325621 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325530 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325521 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325560 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325566 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325625 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325596 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325645 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325511 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325551 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325636 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325622 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325571 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325550 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325583 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325514 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325620 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325556 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325536 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325616 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325586 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325593 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325558 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325630 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325587 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325512 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325561 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325555 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325579 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325606 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325629 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325567 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325518 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325611 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325576 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325634 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325569 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23325510 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03778611 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03986317 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23325547 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23325555 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23325560 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03778624 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03778605 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03221573 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03778623 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03778608 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23325551 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23325581 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23325569 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03221574 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23325583 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23325549 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23325582 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03778606 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23325586 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03327312 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03778617 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03778612 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03778604 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03778629 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23325542 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03778626 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23325567 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23325553 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03610285 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23325543 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23325548 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23325587 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10509015 - -
          Affymetrix Mouse Gene 1.0 ST Array 10509019 - -
          Affymetrix Mouse Gene 1.0 ST Array 10509017 - -
          Affymetrix Mouse Gene 1.0 ST Array 10509020 - -
          Affymetrix Mouse Gene 1.0 ST Array 10509016 - -
          Affymetrix Mouse Gene 1.0 ST Array 10509018 - -
          Affymetrix Mouse Gene 1.0 ST Array 10509022 - -
          Affymetrix Mouse Gene 1.0 ST Array 10509021 - -
          Affymetrix HT MG-430 PM Array Plate 1448538_PM_a_at - -
          Affymetrix HT MG-430 PM Array Plate 1417001_PM_a_at - -
          Affymetrix HT MG-430 PM Array Plate 1435357_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10509014 - -
          Mouse Genome 430 2.0 Array 1448538_PM_a_at - -
          Mouse Genome 430 2.0 Array 1417001_a_at - -
          Mouse Genome 430 2.0 Array 1435357_PM_at - -
          Mouse Genome 430 2.0 Array 1417001_PM_a_at - -
          Mouse Genome 430 2.0 Array 1448538_a_at - -
          Mouse Genome 430 2.0 Array 1435357_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 27981_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 27981_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0400013118.mm.1 NM_023665.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400013118.mm.1 NM_023665.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0400013118.mm.1 TC0400001646.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400013126.mm.1 TC0400001646.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400013120.mm.1 NM_023665.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400013120.mm.1 NM_023665.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0400013120.mm.1 TC0400001646.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400013125.mm.1 TC0400001646.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400013115.mm.1 NM_023665.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400013115.mm.1 NM_023665.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0400013115.mm.1 TC0400001646.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400013123.mm.1 TC0400001646.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400013127.mm.1 TC0400001646.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400013124.mm.1 TC0400001646.mm.1 MTA TC
          Mouse Expression Set 430 1435357_at - -
          Mouse Expression Set 430 1417001_PM_a_at - -
          Mouse Expression Set 430 1435357_PM_at - -
          Mouse Expression Set 430 1448538_a_at - -
          Mouse Expression Set 430 1448538_PM_a_at - -
          Mouse Expression Set 430 1417001_a_at - -
          Murine Genome U74v2 Set 92542_PM_at - -
          Murine Genome U74v2 Set 92542_at - -
          Mouse Genome 430A 2.0 Array 1417001_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1448538_a_at - -
          Mouse Genome 430A 2.0 Array 1448538_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1417001_a_at - -
          Mouse Genome 430A 2.0 Array 1435357_PM_at - -
          Mouse Genome 430A 2.0 Array 1435357_at - -
          Affymetrix HT Mouse Genome 430A Array 1417001_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1435357_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1448538_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1435357_at - -
          Affymetrix HT Mouse Genome 430A Array 1448538_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1417001_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 910735 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 6520114 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006520114 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2731439 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2731439 - -
          mouse-6 v1.1 (Illumina) scl24840.4_10-S - -
          Illumina Sentrix mouse-6.v1 bead array 6520114 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl24840.4_10-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6520114 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2731439 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2731439 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-h9fps:80/100.66.77.141:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline