Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00502731_m1
          See other IGF2BP3 GE Assays ›
          Gene Symbol:
          Igf2bp3
          Gene Name:
          insulin-like growth factor 2 mRNA binding protein 3
          Gene Aliases:
          2610101N11Rik, AA522010, AL022933, AU045931, IMP-3, IMP3, Koc13, Neilsen, mimp3
          Chromosome Location:
          Chr.6: 49085218 - 49215009 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00502731_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Igf2bp3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_023670.3 NP_076159.3 1 - 2 653 63
          GenBank mRNA AB046173.1 - 1 - 2 170 63
          AK011689.1 - 1 - 2 434 63
          AK011797.1 - 1 - 2 404 63
          AK088465.1 - 1 - 2 412 63
          AK147140.1 - 1 - 2 657 63
          BC045138.1 - 1 - 2 283 6306448 63
          BC049082.1 - 1 - 2 319 5706238 63

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600002237.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Igf2bp3

          Panther Classification:

          Molecular Function -

          RNA metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of translation
          transport
          mRNA transport
          nucleotide binding
          nucleic acid binding
          RNA binding
          mRNA 3'-UTR binding
          poly(A) RNA binding
          mRNA 5'-UTR binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0600017585.mm.1 TC0600002237.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600017585.mm.1 NM_023670.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600017585.mm.1 NM_023670.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0600017587.mm.1 TC0600002237.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600017587.mm.1 NM_023670.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600017587.mm.1 NM_023670.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0600017586.mm.1 TC0600002237.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600017586.mm.1 NM_023670.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600017586.mm.1 NM_023670.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0600017588.mm.1 TC0600002237.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600017588.mm.1 NM_023670.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600017588.mm.1 NM_023670.3 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 626722 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0