Search
Search

Wdr54| Gene Symbol: Wdr54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023790.2 | NP_076279.1 | 6 - 7 | 743 | 82 | |
| GenBank mRNA | AF053628.1 | - | 6 - 7 | 731 | 82 | |
| AK006536.1 | - | 6 - 7 | 643 | 82 | ||
| AK154345.1 | - | 6 - 7 | 734 | 82 | ||
| BC083332.1 | - | 7 - 8 | 648 | 6819838 | 82 | |
Set Membership: |
|