Search
Search

Tmem176a| Gene Symbol: Tmem176a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098271.1 | NP_001091741.1 | 3 - 4 | 364 | 110 | |
| NM_025326.4 | NP_079602.4 | 3 - 4 | 416 | 110 | ||
| GenBank mRNA | AB063313.1 | - | 2 - 3 | 306 | 110 | |
| AK002546.1 | - | 3 - 4 | 392 | 110 | ||
| AK004315.1 | - | 3 - 4 | 389 | 110 | ||
| AK079378.1 | - | 378 | 110 | |||
| AK150576.1 | - | 3 - 4 | 381 | 110 | ||
| BC010831.1 | - | 3 - 4 | 360 | 4221626 | 110 | |
| CT010190.1 | - | 2 - 3 | 306 | 110 | ||
Set Membership: |
|