Search
Search

Myo19| Gene Symbol: Myo19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025414.3 | NP_079690.2 | 19 - 20 | 2398 | 80 | |
| GenBank mRNA | AK018609.1 | - | 4 - 5 | 428 | 80 | |
| AK080117.1 | - | 17 - 18 | 2117 | 80 | ||
| BC007156.1 | - | 12 - 13 | 1314 | 3486558 | 80 | |
| BC046294.1 | - | 18 - 19 | 2126 | 80 | ||
| BC060115.1 | - | 17 - 18 | 2251 | 80 | ||
Set Membership: |
|