Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00502977_m1
          See other NICN1 GE Assays ›
          Gene Symbol:
          Nicn1
          Gene Name:
          nicolin 1
          Gene Aliases:
          1500032A17Rik
          Chromosome Location:
          Chr.9: 108290457 - 108296496 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00502977_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nicn1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_025449.3 NP_079725.1 1 - 2 207 86
          GenBank mRNA AJ299741.1 - 1 - 2 199 86
          AK002789.1 - 1 - 2 213 86
          AK013602.1 - 1 - 2 235 86
          AK075732.1 - 1 - 2 215 86
          AK146890.1 - 1 - 2 217 86
          AK169135.1 - 1 - 2 217 86
          BC024050.1 - 1 - 2 207 5133763 86

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0900001398.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Nicn1

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P94845 BC024050.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P94845 AK075732.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P94845 AK146890.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P94845 AK169135.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P94845 NM_025449.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P94845 BC024050.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P94845 AK075732.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P94845 AK146890.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P94845 AK169135.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P94845 NM_025449.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 843077 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 89117 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 461352 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 26185 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P166513 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P94845 BC024050.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P94845 AK075732.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P94845 AK146890.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P94845 AK169135.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P94845 NM_025449.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P94845 BC024050.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P94845 AK075732.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P94845 AK146890.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P94845 AK169135.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P94845 NM_025449.3 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26814082 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26814101 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26814070 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26814090 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26814112 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26814120 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26814092 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26814088 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26814119 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26814075 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26814068 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26814103 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26814079 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26814094 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26814086 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26814069 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26814113 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26814108 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26814098 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26814111 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26814078 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26814074 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26814107 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26814089 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26814072 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26814099 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26814106 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26814109 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26814105 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26814117 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26814122 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26814114 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26814102 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P166513 - -
          Whole Mouse Genome G4122A A_52_P94845 BC024050.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P94845 AK075732.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P94845 AK146890.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P94845 AK169135.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P94845 NM_025449.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P94845 BC024050.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P94845 AK075732.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P94845 AK146890.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P94845 AK169135.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P94845 NM_025449.3 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P118445 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P166513 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07879367 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07597528 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07594231 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07879373 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07597523 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07879374 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07597529 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07879371 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814101 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814155 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814124 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814185 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814170 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814126 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814128 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814162 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814111 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814191 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814086 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814119 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814107 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814151 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814120 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814140 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814099 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814181 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814130 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814075 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814141 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814165 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814157 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814102 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814074 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814195 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814112 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814174 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814105 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814122 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814145 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814081 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814176 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814187 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814129 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814114 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814068 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814088 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814193 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814103 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814131 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814148 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814104 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814117 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814190 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814109 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814118 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814160 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814090 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814113 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814179 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814100 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814197 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814065 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814070 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26814168 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26814120 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07773570 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26814075 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08041391 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07866519 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07773572 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26814112 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26814072 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26814078 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07866518 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26814122 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26814119 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26814089 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26814074 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26814070 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26814105 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26814103 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26814117 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26814094 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26814111 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26814081 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07879372 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26814088 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26814079 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26814067 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26814073 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26814114 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26814109 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07879365 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26814092 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26814107 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07594231 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26814068 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10588882 - -
          Affymetrix Mouse Gene 1.0 ST Array 10588878 - -
          Affymetrix Mouse Gene 1.0 ST Array 10588880 - -
          Affymetrix Mouse Gene 1.0 ST Array 10588881 - -
          Affymetrix Mouse Gene 1.0 ST Array 10588879 - -
          Affymetrix Mouse Gene 1.0 ST Array 10588877 - -
          Affymetrix HT MG-430 PM Array Plate 1449376_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1449377_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10588876 - -
          Mouse Genome 430 2.0 Array 1449377_at - -
          Mouse Genome 430 2.0 Array 1449376_PM_at - -
          Mouse Genome 430 2.0 Array 1449377_PM_at - -
          Mouse Genome 430 2.0 Array 1449376_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66257_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66257_at - -
          Affymetrix Mouse Clariom S Array PSR0900011209.mm.1 NM_025449.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900011209.mm.1 NM_025449.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0900011209.mm.1 TC0900001398.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900011210.mm.1 NM_025449.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900011210.mm.1 NM_025449.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0900011210.mm.1 TC0900001398.mm.1 MTA TC
          Mouse Expression Set 430 1449377_at - -
          Mouse Expression Set 430 1449376_PM_at - -
          Mouse Expression Set 430 1449377_PM_at - -
          Mouse Expression Set 430 1449376_at - -
          Murine Genome U74v2 Set 102059_at - -
          Murine Genome U74v2 Set 102059_PM_at - -
          Mouse Genome 430A 2.0 Array 1449376_PM_at - -
          Mouse Genome 430A 2.0 Array 1449376_at - -
          Mouse Genome 430A 2.0 Array 1449377_PM_at - -
          Mouse Genome 430A 2.0 Array 1449377_at - -
          Affymetrix HT Mouse Genome 430A Array 1449376_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1449377_at - -
          Affymetrix HT Mouse Genome 430A Array 1449377_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1449376_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 418918 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 4050403 - -
          Illumina mouse-6 v1.0 expression beadchip 4050403 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004050403 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2594394 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2594394 - -
          mouse-6 v1.1 (Illumina) scl36477.6_179-S - -
          Illumina Sentrix mouse-6.v1 bead array 4050403 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl36477.6_179-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4050403 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2821916 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2594394 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2594394 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2821916 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl36477.6_179-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline