| Array Name |
Probe IDs |
Targets |
Target Type |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P397564 |
AK002371.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P397564 |
AK002371.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2185203 |
BC058365.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2185203 |
AK148443.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2185203 |
AK152287.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2185203 |
AK152352.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2185203 |
AK169605.1 |
mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
319066 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
876553 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
719959 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
612946 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
359575 |
AK148411.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
359575 |
AK165677.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
359575 |
BC022914.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
359575 |
BC027061.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
359575 |
NM_183165.3 |
refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
554128 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
94642 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
602820 |
AK048911.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
186028 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
568311 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
973163 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
591031 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
958792 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
694958 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
821601 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
780490 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
933083 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
165893 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
124176 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
81045 |
- |
- |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P397564 |
AK002371.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P397564 |
AK002371.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2185203 |
BC058365.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2185203 |
AK148443.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2185203 |
AK152287.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2185203 |
AK152352.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2185203 |
AK169605.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P397564 |
AK002371.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P397564 |
AK002371.1 |
mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P119927 |
- |
- |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P105652 |
- |
- |