Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00503102_m1
          See other MED28 GE Assays ›
          Gene Symbol:
          Med28
          Gene Name:
          mediator complex subunit 28
          Gene Aliases:
          1500003D12Rik, AI451633, AU045690, Eg1, FKSG20, magicin
          Chromosome Location:
          Chr.5: 45520229 - 45529284 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00503102_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Med28
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_025895.4 NP_080171.1 1 - 2 259 70
          GenBank mRNA AF318060.1 - 1 - 2 183 70
          AK003854.1 - 1 - 2 172 70
          AK005681.1 - 1 - 2 168 70
          AK007855.1 - 1 - 2 188 70
          AK154249.1 - 1 - 2 185 70
          BC024941.1 - 1 - 2 202 4017146 70
          BY725533.1 - 1 - 2 239 70
          DV653139.1 - 1 - 2 259 70

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500000489.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Med28

          Panther Classification:

          Molecular Function -

          general transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          stem cell population maintenance
          negative regulation of smooth muscle cell differentiation
          actin binding
          protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23657076 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23657077 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23657074 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23657072 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23657073 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2066329 NM_025895.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_65_P01783 AK154249.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_65_P01783 BC024941.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_65_P01783 BY725533.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_65_P01783 DV653139.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_65_P01783 AK005681.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1956712 AK154249.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1956712 AK005130.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1956712 AK005681.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1956712 AK007855.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1956712 NM_025895.4 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 633044 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 376959 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 244376 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 215535 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 225493 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 907607 AK011300.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 907607 AK015403.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 907607 AK082082.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 907607 AK082331.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 907607 AK129130.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 907607 BC021794.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 907607 NM_026440.3 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 39733 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P133563 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P100175 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P141883 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2066329 NM_025895.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1956712 AK154249.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1956712 AK005130.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1956712 AK005681.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1956712 AK007855.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1956712 NM_025895.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_65_P01783 AK154249.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_65_P01783 BC024941.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_65_P01783 BY725533.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_65_P01783 DV653139.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_65_P01783 AK005681.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23657059 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23657082 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23657072 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23657077 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23657081 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23657064 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23657061 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23657071 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23657088 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23657078 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23657075 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23657068 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23657091 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23657065 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23657074 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23657073 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23657056 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23657089 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23657086 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23657084 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23657066 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23657079 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23657093 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P133563 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P100175 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P141883 - -
          Whole Mouse Genome G4122A A_52_P552580 AK154249.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P552580 BC024941.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P552580 AK005130.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P552580 AK005681.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P552580 AK007855.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P362245 AK154249.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P362245 AK005130.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P362245 NM_025895.4 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P125001 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P141883 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P100175 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P133563 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04860645 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04967487 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04860649 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04860643 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23657074 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23657073 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23657077 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23657072 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23657076 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23657075 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23657078 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23657042 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23657128 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23657061 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23657084 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23657103 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23657072 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23657047 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23657089 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23657071 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23657116 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23657110 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23657043 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23657088 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23657045 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23657082 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23657086 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23657136 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23657064 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23657120 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23657127 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23657051 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23657102 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23657113 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23657065 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23657130 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23657040 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23657095 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23657039 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23657038 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23657115 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23657052 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23657105 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23657079 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23657112 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23657056 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23657074 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23657073 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23657133 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23657049 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23657123 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23657093 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23657099 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23657122 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23657091 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23657081 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23657111 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23657137 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23657086 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23657120 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23657049 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23657071 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23657082 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04835418 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23657057 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23657123 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04195439 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04203014 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04465386 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23657043 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23657061 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23657060 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04847181 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23657068 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04387161 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23657072 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23657054 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23657096 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23657056 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23657116 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23657095 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04933892 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23657042 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23657047 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23657110 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23657052 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23657081 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23657078 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04387160 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23657112 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23657074 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23657115 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23657091 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23657066 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04844774 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23657111 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04844776 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23657040 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23657113 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04598072 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23657105 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23657051 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23657093 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23657084 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04860643 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23657088 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04603840 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04844773 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23657065 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23657077 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10521730 - -
          Affymetrix Mouse Gene 1.0 ST Array 10521726 - -
          Affymetrix Mouse Gene 1.0 ST Array 10521727 - -
          Affymetrix Mouse Gene 1.0 ST Array 10521729 - -
          Affymetrix Mouse Gene 1.0 ST Array 10521728 - -
          Affymetrix HT MG-430 PM Array Plate 1448374_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1416508_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10521725 - -
          Mouse Genome 430 2.0 Array 1448374_at - -
          Mouse Genome 430 2.0 Array 1416508_PM_at - -
          Mouse Genome 430 2.0 Array 1448374_PM_at - -
          Mouse Genome 430 2.0 Array 1416508_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66999_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66999_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0500004614.mm.1 TC0500000489.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500004614.mm.1 NM_025895.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500004614.mm.1 NM_025895.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0500004608.mm.1 TC0500000489.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500004608.mm.1 NM_025895.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500004608.mm.1 NM_025895.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0500004611.mm.1 TC0500000489.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500004611.mm.1 NM_025895.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500004611.mm.1 NM_025895.4 Refseq
          Mouse Expression Set 430 1416508_PM_at - -
          Mouse Expression Set 430 1448374_PM_at - -
          Mouse Expression Set 430 1416508_at - -
          Mouse Expression Set 430 1448374_at - -
          Murine Genome U74v2 Set 95044_PM_at - -
          Murine Genome U74v2 Set 95044_at - -
          Mouse Genome 430A 2.0 Array 1448374_PM_at - -
          Mouse Genome 430A 2.0 Array 1416508_at - -
          Mouse Genome 430A 2.0 Array 1416508_PM_at - -
          Mouse Genome 430A 2.0 Array 1448374_at - -
          Affymetrix HT Mouse Genome 430A Array 1448374_at - -
          Affymetrix HT Mouse Genome 430A Array 1416508_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1448374_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1416508_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 854180 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 2570131 - -
          Illumina mouse-6 v1.0 expression beadchip 101500167 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002570131 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2637365 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2637365 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1256361 - -
          mouse-6 v1.1 (Illumina) scl27845.5_85-S - -
          mouse-6 v1.1 (Illumina) scl0066999.2_129-S - -
          Illumina Sentrix mouse-6.v1 bead array 101500167 - -
          Illumina Sentrix mouse-6.v1 bead array 2570131 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0066999.2_129-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 101500167 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2570131 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2637365 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2637365 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0066999.2_129-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline