Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00503220_m1
          See other GADD45GIP1 GE Assays ›
          Gene Symbol:
          Gadd45gip1
          Gene Name:
          growth arrest and DNA-damage-inducible, gamma interacting protein 1
          Gene Aliases:
          2310040G17Rik, AI425883, Crif1, MRP-L59, Plinp1
          Chromosome Location:
          Chr.8: 84832282 - 84835482 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00503220_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gadd45gip1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_183358.4 NP_899202.3 1 - 2 362 97
          GenBank mRNA AK002738.1 - 1 - 2 368 97
          AK003314.1 - 1 - 2 363 97
          AK009723.1 - 1 - 2 361 97
          AK019570.1 - 1 - 2 363 97
          AK028227.1 - 1 - 2 363 97
          BC022592.1 - 1 - 2 247 97
          BC061069.1 - 1 - 2 362 6740575 97

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0800000962.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gadd45gip1

          Gene Ontology Categories:

          Function(s)

          cell cycle
          mitotic cell cycle arrest

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26022162 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26022164 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26022163 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2150633 AK003314.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2150633 AK009723.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2150633 AK019570.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2150633 AK028227.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2150633 BC022592.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P85020 AK028227.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P85020 NM_183358.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P85020 NM_009010.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P85020 AK038300.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P85020 AK028227.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P85020 NM_183358.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P85020 NM_009010.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P85020 AK038300.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 250272 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 591284 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 155982 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P179210 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2150633 AK003314.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2150633 AK009723.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2150633 AK019570.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2150633 AK028227.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2150633 BC022592.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P85020 AK028227.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P85020 NM_183358.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P85020 NM_009010.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P85020 AK038300.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P85020 AK028227.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P85020 NM_183358.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P85020 NM_009010.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P85020 AK038300.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26022163 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26022168 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26022164 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26022169 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26022162 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26022161 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26022173 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26022172 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26022174 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26022166 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26022176 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P179210 - -
          Whole Mouse Genome G4122A A_51_P419086 AK003314.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P419086 AK009723.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P419086 AK028227.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P419086 NM_183358.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P85020 AK028227.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P85020 NM_183358.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P85020 NM_009010.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P85020 AK038300.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P85020 AK028227.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P85020 NM_183358.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P85020 NM_009010.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P85020 AK038300.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P114043 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P179210 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06809817 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26022163 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26022162 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26022164 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022238 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022165 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022195 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022178 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022247 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022141 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022130 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022127 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022151 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022224 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022204 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022133 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022227 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022197 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022200 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022210 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022138 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022220 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022199 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022153 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022181 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022134 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022257 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022233 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022159 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022143 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022172 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022168 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022158 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022229 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022244 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022252 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022260 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022205 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022249 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022163 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022192 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022129 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022169 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022164 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022245 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022179 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022231 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022156 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022161 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022146 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022182 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022235 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022207 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022211 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022258 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022160 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022152 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022166 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022217 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022255 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022176 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022170 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022214 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022228 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022202 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022174 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07342016 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022179 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022168 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022176 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022161 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022152 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022170 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07128369 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022159 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022163 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022156 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06830264 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022166 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022164 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022174 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022182 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06814524 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022153 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022181 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022178 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022169 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022162 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022149 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022165 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022151 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022160 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10573432 - -
          Affymetrix Mouse Gene 1.0 ST Array 10573431 - -
          Affymetrix Mouse Gene 1.0 ST Array 10573433 - -
          Affymetrix HT MG-430 PM Array Plate 1448776_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1417619_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1446783_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10580210 - -
          Affymetrix Mouse Gene 1.1 ST Array 10573430 - -
          Mouse Genome 430 2.0 Array 1446783_at - -
          Mouse Genome 430 2.0 Array 1417619_PM_at - -
          Mouse Genome 430 2.0 Array 1448776_at - -
          Mouse Genome 430 2.0 Array 1417619_at - -
          Mouse Genome 430 2.0 Array 1446783_PM_at - -
          Mouse Genome 430 2.0 Array 1448776_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 102060_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 102060_at - -
          Affymetrix Mouse Clariom S Array PSR0800007269.mm.1 NM_183358.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800007269.mm.1 NM_183358.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0800007269.mm.1 TC0800000962.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800007267.mm.1 NM_183358.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800007267.mm.1 NM_183358.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0800007267.mm.1 TC0800000962.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800007268.mm.1 NM_183358.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800007268.mm.1 NM_183358.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0800007268.mm.1 TC0800000962.mm.1 MTA TC
          Mouse Expression Set 430 1448776_at - -
          Mouse Expression Set 430 1417619_at - -
          Mouse Expression Set 430 1417619_PM_at - -
          Mouse Expression Set 430 1448776_PM_at - -
          Mouse Expression Set 430 1446783_PM_at - -
          Mouse Expression Set 430 1446783_at - -
          Mouse Genome 430A 2.0 Array 1448776_at - -
          Mouse Genome 430A 2.0 Array 1448776_PM_at - -
          Mouse Genome 430A 2.0 Array 1417619_at - -
          Mouse Genome 430A 2.0 Array 1417619_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1417619_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1448776_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1448776_at - -
          Affymetrix HT Mouse Genome 430A Array 1417619_at - -
          Affymetrix HT Mouse Genome 430B Array 1446783_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1446783_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 921350 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2970047 - -
          Illumina mouse-6 v1.0 expression beadchip 2970047 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002970047 - -
          mouse-6 v1.1 (Illumina) scl33577.2_419-S - -
          Illumina Sentrix mouse-6.v1 bead array 2970047 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl33577.2_419-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2970047 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2830860 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2830860 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1240677 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl33577.2_419-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline