Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00503305_g1
          See other TSR3 GE Assays ›
          Gene Symbol:
          Tsr3
          Gene Name:
          TSR3 20S rRNA accumulation
          Gene Aliases:
          0610007P22Rik, 1110014B11Rik, AL022759
          Chromosome Location:
          Chr.17: 25240170 - 25242799 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00503305_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tsr3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001163718.1 NP_001157190.1 1 - 2 198 118
          NM_026676.3 NP_080952.2 1 - 2 198 118
          GenBank mRNA AK002309.1 - 1 - 2 170 118
          AK158743.1 - 1 - 2 199 118
          AK169072.1 - 1 - 2 133 118
          AK170074.1 - 1 - 2 167 118
          BC027751.1 - 1 - 2 203 5011894 118
          BC089019.1 - 1 - 2 147 30551363 118
          CA979987.1 - 1 - 2 147 118

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700000367.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the BC027751.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tsr3

          Gene Ontology Categories:

          Function(s) Process(es)

          rRNA processing
          biological_process
          maturation of SSU-rRNA
          ribosome biogenesis
          molecular_function

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1700003041.mm.1 NM_001163718.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700003041.mm.1 NM_001163718.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700003041.mm.1 NM_026676.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700003041.mm.1 NM_026676.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700003041.mm.1 TC1700000367.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700003043.mm.1 NM_001163718.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700003043.mm.1 NM_001163718.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700003043.mm.1 NM_026676.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700003043.mm.1 NM_026676.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700003043.mm.1 TC1700000367.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700003044.mm.1 NM_001163718.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700003044.mm.1 NM_001163718.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700003044.mm.1 NM_026676.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700003044.mm.1 NM_026676.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700003044.mm.1 TC1700000367.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700003048.mm.1 TC1700000367.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 580017 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline