Search
Search

Aspdh| Gene Symbol: Aspdh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026690.1 | NP_080966.1 | 2 - 3 | 245 | 61 | |
| GenBank mRNA | AK002587.1 | - | 2 - 3 | 245 | 61 | |
| BC019451.1 | - | 2 - 3 | 211 | 4194890 | 61 | |
Set Membership: |
|