Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00503356_g1
          See other TLCD1 GE Assays ›
          Gene Symbol:
          Tlcd1
          Gene Name:
          TLC domain containing 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.11: 78176716 - 78180820 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00503356_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tlcd1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001291235.1 NP_001278164.1 3 - 4 250 89
          NM_001291236.1 NP_001278165.1 3 - 4 726 89
          NM_001291237.1 NP_001278166.1 4 - 5 434 89
          NM_026708.2 NP_080984.1 3 - 4 561 89
          GenBank mRNA AK046577.1 - 4 - 5 2441 89
          AK161721.1 - 461 89
          AK165432.1 - 673 89
          BC005702.1 - 3 - 4 453 3598444 89
          BC039182.1 - 3 - 4 696 3500908 89
          BG083251.2 - 3 - 4 308 89

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100001147.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tlcd1

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27941791 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27941790 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27941789 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2108599 BC005702.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2108599 BC039182.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2108599 AK161721.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2108599 AK165432.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2108599 AK002703.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 380474 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 398939 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P109624 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2108599 BC005702.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2108599 BC039182.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2108599 AK161721.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2108599 AK165432.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2108599 AK002703.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27941800 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27941795 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27941796 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27941792 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27941791 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27941805 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27941789 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27941803 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27941794 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27941798 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27941804 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27941802 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P109624 - -
          Whole Mouse Genome G4122A A_52_P611650 AK002703.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P312275 BC005702.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P312275 BC039182.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P312275 AK161721.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P312275 AK165432.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P312275 AK002265.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P331215 AK161721.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P133315 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P109624 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09457746 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09457730 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09457731 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27941791 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27941790 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27941789 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27941791 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27941749 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27941851 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27941798 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27941800 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27941782 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27941734 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27941863 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27941753 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27941861 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27941803 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27941805 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27941829 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27941776 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27941845 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27941726 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27941862 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27941768 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27941720 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27941792 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27941840 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27941855 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27941721 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27941842 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27941816 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27941747 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27941738 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27941817 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27941780 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27941787 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27941802 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27941723 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27941761 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27941770 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27941853 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27941859 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27941795 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27941796 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27941848 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27941850 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27941728 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27941827 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27941778 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27941763 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27941775 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27941825 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27941736 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27941760 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27941813 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27941786 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27941804 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27941758 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27941785 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27941830 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27941789 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27941856 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27941783 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27941773 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27941751 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27941791 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27941789 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27941795 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09457743 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09457755 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27941798 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27941802 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27941800 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27941792 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09307412 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27941803 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27941785 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27941804 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27941783 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27941786 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10379035 - -
          Affymetrix Mouse Gene 1.0 ST Array 10379039 - -
          Affymetrix Mouse Gene 1.0 ST Array 10379041 - -
          Affymetrix Mouse Gene 1.0 ST Array 10379043 - -
          Affymetrix Mouse Gene 1.0 ST Array 10379038 - -
          Affymetrix Mouse Gene 1.0 ST Array 10379040 - -
          Affymetrix Mouse Gene 1.0 ST Array 10379037 - -
          Affymetrix Mouse Gene 1.0 ST Array 10379042 - -
          Affymetrix HT MG-430 PM Array Plate 1452132_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1426616_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1436404_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1430084_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10379034 - -
          Mouse Genome 430 2.0 Array 1452132_at - -
          Mouse Genome 430 2.0 Array 1430084_at - -
          Mouse Genome 430 2.0 Array 1430084_PM_at - -
          Mouse Genome 430 2.0 Array 1436404_PM_at - -
          Mouse Genome 430 2.0 Array 1452132_PM_at - -
          Mouse Genome 430 2.0 Array 1436404_at - -
          Mouse Genome 430 2.0 Array 1426616_PM_at - -
          Mouse Genome 430 2.0 Array 1426616_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 68385_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 68385_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1100010538.mm.1 NM_026708.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100010538.mm.1 TC1100001147.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100010540.mm.1 NM_026708.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100010540.mm.1 TC1100001147.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100010542.mm.1 NM_026708.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100010542.mm.1 TC1100001147.mm.1 MTA TC
          Mouse Expression Set 430 1436404_at - -
          Mouse Expression Set 430 1436404_PM_at - -
          Mouse Expression Set 430 1426616_at - -
          Mouse Expression Set 430 1452132_at - -
          Mouse Expression Set 430 1452132_PM_at - -
          Mouse Expression Set 430 1426616_PM_at - -
          Mouse Expression Set 430 1430084_PM_at - -
          Mouse Expression Set 430 1430084_at - -
          Mouse Genome 430A 2.0 Array 1426616_PM_at - -
          Mouse Genome 430A 2.0 Array 1426616_at - -
          Mouse Genome 430A 2.0 Array 1452132_at - -
          Mouse Genome 430A 2.0 Array 1436404_at - -
          Mouse Genome 430A 2.0 Array 1436404_PM_at - -
          Mouse Genome 430A 2.0 Array 1452132_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1436404_at - -
          Affymetrix HT Mouse Genome 430A Array 1452132_at - -
          Affymetrix HT Mouse Genome 430A Array 1452132_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1426616_at - -
          Affymetrix HT Mouse Genome 430A Array 1436404_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1426616_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1430084_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1430084_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 106550364 - -
          Illumina mouse-6 v1.0 expression beadchip 106550364 - -
          Illumina mouse-6 v1.0 expression beadchip 6840438 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006840438 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2624326 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2624326 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1253221 - -
          mouse-6 v1.1 (Illumina) scl068385.4_14-S - -
          Illumina Sentrix mouse-6.v1 bead array 6840438 - -
          Illumina Sentrix mouse-6.v1 bead array 106550364 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl068385.4_14-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 106550364 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6840438 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2781458 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2781458 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1253221 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl068385.4_14-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-j77tp:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline