Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00503373_m1
          See other ICT1 GE Assays ›
          Gene Symbol:
          Ict1
          Gene Name:
          immature colon carcinoma transcript 1
          Gene Aliases:
          1110001A02Rik, 1110002E03Rik, MRP-L58
          Chromosome Location:
          Chr.11: 115403725 - 115410919 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00503373_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ict1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_026729.1 NP_081005.1 1 - 2 108 95
          GenBank mRNA AK003192.1 - 1 - 2 108 95

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100001892.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ict1

          Panther Classification:

          Molecular Function -

          translation factor translation release factor translational protein

          Gene Ontology Categories:

          Function(s) Process(es)

          translation
          translational termination
          mitochondrial translational termination
          translation release factor activity
          aminoacyl-tRNA hydrolase activity
          translation release factor activity, codon nonspecific
          hydrolase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28153292 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28153288 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28153346 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28153347 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28153290 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28153286 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28153289 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28153287 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1974134 AK160697.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1974134 BC028523.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1974134 BC079614.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1974134 NM_026729.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1974134 AK003192.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 141659 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 696909 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 763855 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 48300 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 941738 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P127943 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1974134 AK160697.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1974134 BC028523.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1974134 BC079614.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1974134 NM_026729.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1974134 AK003192.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28153269 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28153296 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28153266 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28153265 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28153259 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28153280 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28153284 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28153278 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28153261 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28153290 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28153282 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28153303 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28153289 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28153267 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28153274 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28153294 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28153277 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28153297 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28153292 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28153301 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28153271 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28153300 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28153263 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28153288 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P127943 - -
          Whole Mouse Genome G4122A A_51_P424999 AK160697.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P424999 BC028523.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P424999 BC079614.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P424999 NM_026729.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P424999 AK003192.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P110663 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P127950 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P127943 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09212757 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09649947 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28153287 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28153347 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28153288 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28153346 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28153286 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28153292 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28153290 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28153289 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153312 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153396 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153306 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153289 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153352 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153354 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153373 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153334 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153266 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153307 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153386 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153281 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153359 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153310 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153350 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153379 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153365 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153349 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153341 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153249 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153330 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153255 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153263 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153326 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153353 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153351 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153394 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153382 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153264 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153378 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153267 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153393 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153328 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153324 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153325 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153389 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153344 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153366 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153265 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153338 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153339 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153301 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153395 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153286 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153369 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153278 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153288 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153391 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153271 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153274 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153284 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153315 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153388 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153364 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153347 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153290 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153384 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153282 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153309 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153311 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153377 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153397 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153261 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153332 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153367 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153361 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153269 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153398 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153323 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153253 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153360 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153259 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153346 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153277 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153300 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153348 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153370 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153371 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153292 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153355 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28153284 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28153271 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28153312 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28153254 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28153335 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28153277 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28153289 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09547511 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28153282 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28153259 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28153347 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09382542 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09237037 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28153341 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28153249 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09315533 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09308137 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28153328 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28153315 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28153344 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28153281 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28153317 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28153324 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09414866 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28153294 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28153321 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09680321 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28153330 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09700350 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28153267 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28153299 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09139501 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08981078 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28153346 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28153303 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09139504 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28153263 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28153274 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28153290 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28153301 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28153269 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28153304 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28153296 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28153326 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09315534 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09237038 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28153261 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28153332 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28153325 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28153255 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09649947 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09537818 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28153264 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28153292 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09308136 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09382544 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28153339 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28153278 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28153334 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28153310 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28153311 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09212757 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28153251 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10382509 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382515 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382512 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382511 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382513 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382514 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382510 - -
          Affymetrix HT MG-430 PM Array Plate 1460308_PM_a_at - -
          Affymetrix HT MG-430 PM Array Plate 1444276_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10382508 - -
          Mouse Genome 430 2.0 Array 1460308_PM_a_at - -
          Mouse Genome 430 2.0 Array 1444276_at - -
          Mouse Genome 430 2.0 Array 1460308_a_at - -
          Mouse Genome 430 2.0 Array 1444276_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 68572_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 68572_at - -
          Affymetrix Mouse Clariom S Array PSR1100017760.mm.1 NM_026729.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100017760.mm.1 NM_026729.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100017760.mm.1 TC1100001892.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100017761.mm.1 NM_026729.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100017761.mm.1 NM_026729.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100017761.mm.1 TC1100001892.mm.1 MTA TC
          Mouse Expression Set 430 1460308_a_at - -
          Mouse Expression Set 430 1460308_PM_a_at - -
          Mouse Expression Set 430 1444276_at - -
          Mouse Expression Set 430 1444276_PM_at - -
          Murine Genome U74v2 Set 99150_at - -
          Murine Genome U74v2 Set 99150_PM_at - -
          Mouse Genome 430A 2.0 Array 1460308_a_at - -
          Mouse Genome 430A 2.0 Array 1460308_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1460308_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1460308_PM_a_at - -
          Affymetrix HT Mouse Genome 430B Array 1444276_at - -
          Affymetrix HT Mouse Genome 430B Array 1444276_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 376401 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 430138 - -
          Illumina mouse-6 v1.0 expression beadchip 430138 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000430138 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2642063 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2642063 - -
          mouse-6 v1.1 (Illumina) scl40732.5.1_61-S - -
          Illumina Sentrix mouse-6.v1 bead array 430138 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl40732.5.1_61-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 430138 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2642063 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2642063 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl40732.5.1_61-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline