Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00503412_m1
          See other 2310036O22RIK GE Assays ›
          Gene Symbol:
          2310036O22Rik
          Gene Name:
          RIKEN cDNA 2310036O22 gene
          Gene Aliases:
          1110002H14Rik, AI854688
          Chromosome Location:
          Chr.8: 85026814 - 85030286 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00503412_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 2310036O22Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_026760.2 NP_081036.2 1 - 2 406 93
          GenBank mRNA AK003299.1 - 1 - 2 371 93
          AK009653.1 - 1 - 2 373 93
          BC023284.1 - 1 - 2 416 3484935 93
          BC029622.1 - 1 - 2 406 4207296 93

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0800000976.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene 2310036O22Rik

          Panther Classification:

          Molecular Function -

          DNA metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26023271 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26023270 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26023269 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P286321 BC029622.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P286321 BC023284.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P286321 NM_026760.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P286321 AK003299.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P286321 AK009653.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P286321 BC029622.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P286321 BC023284.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P286321 NM_026760.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P286321 AK003299.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P286321 AK009653.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 743765 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 85848 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P131992 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P286321 BC029622.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P286321 BC023284.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P286321 NM_026760.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P286321 AK003299.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P286321 AK009653.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P286321 BC029622.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P286321 BC023284.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P286321 NM_026760.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P286321 AK003299.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P286321 AK009653.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26023278 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26023276 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26023273 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26023274 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26023288 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26023283 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26023287 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26023281 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26023289 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26023291 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26023272 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26023285 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26023282 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26023271 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26023284 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26023269 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26023290 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P131992 - -
          Whole Mouse Genome G4122A A_51_P286321 BC029622.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P286321 BC023284.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P286321 NM_026760.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P286321 AK003299.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P286321 AK009653.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P286321 BC029622.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P286321 BC023284.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P286321 NM_026760.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P286321 AK003299.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P286321 AK009653.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P119689 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P131992 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07140101 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26023271 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26023269 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26023270 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26023315 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26023321 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26023281 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26023218 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26023279 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26023223 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26023349 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26023269 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26023266 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26023237 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26023333 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26023342 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26023291 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26023273 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26023203 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26023339 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26023265 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26023227 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26023348 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26023335 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26023220 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26023205 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26023309 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26023232 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26023206 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26023277 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26023271 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26023274 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26023332 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26023207 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26023288 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26023212 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26023217 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26023211 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26023255 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26023307 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26023312 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26023248 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26023322 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26023287 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26023350 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26023233 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26023210 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26023282 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26023330 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26023343 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26023318 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26023253 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26023200 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26023317 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26023250 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26023324 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26023234 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26023326 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26023345 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26023221 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26023285 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26023259 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26023283 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26023215 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26023252 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26023267 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26023305 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26023257 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26023327 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26023336 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26023261 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26023283 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07418749 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26023266 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26023277 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26023282 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26023264 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26023269 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07046619 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26023285 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26023281 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26023276 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26023279 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26023289 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07140102 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26023287 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26023263 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26023288 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26023265 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10573516 - -
          Affymetrix Mouse Gene 1.0 ST Array 10573517 - -
          Affymetrix Mouse Gene 1.0 ST Array 10573518 - -
          Affymetrix HT MG-430 PM Array Plate 1428201_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10573515 - -
          Mouse Genome 430 2.0 Array 1428201_PM_at - -
          Mouse Genome 430 2.0 Array 1428201_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 68544_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 68544_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0800007453.mm.1 NM_026760.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800007453.mm.1 NM_026760.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800007453.mm.1 TC0800000976.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800007452.mm.1 NM_026760.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800007452.mm.1 NM_026760.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800007452.mm.1 TC0800000976.mm.1 MTA TC
          Affymetrix HT Mouse Genome 430A Array 1428201_at - -
          Affymetrix HT Mouse Genome 430A Array 1428201_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 932994 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 4560239 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004560239 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1229716 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1229716 - -
          mouse-6 v1.1 (Illumina) scl068544.3_98-S - -
          Illumina Sentrix mouse-6.v1 bead array 4560239 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl068544.3_98-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4560239 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1229716 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1229716 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl068544.3_98-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline