Search
Search

Rarres2| Gene Symbol: Rarres2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027852.2 | NP_082128.1 | 3 - 4 | 379 | 74 | |
| GenBank mRNA | AK002298.1 | - | 3 - 4 | 380 | 74 | |
| BC038914.1 | - | 3 - 4 | 399 | 5100583 | 74 | |
Set Membership: |
|