Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00503985_m1
          See other SPATA19 GE Assays ›
          Gene Symbol:
          Spata19
          Gene Name:
          spermatogenesis associated 19
          Gene Aliases:
          1700001L23Rik, AV047838, spergen-1
          Chromosome Location:
          Chr.9: 27396821 - 27401711 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00503985_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Spata19
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AB120716.1 - 4 - 5 373 71
          AK005610.1 - 4 - 5 373 71
          BC049742.1 - 3 - 4 317 6771761 71
          JX089334.1 - 4 - 5 673 71

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0900000261.mm.1

          Set Membership:

          Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2017535 AB120716.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2017535 AK005610.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2017535 NM_029299.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2067246 BC049742.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 735334 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 112203 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 564842 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 61665 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P111427 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2017535 AB120716.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2017535 AK005610.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2017535 NM_029299.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26375855 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26375854 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26375869 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26375861 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26375860 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26375851 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26375852 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26375862 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26375866 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26375846 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26375856 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26375867 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26375847 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26375870 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26375865 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26375849 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26375844 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26375858 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26375845 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P111427 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01890947 - -
          Whole Mouse Genome G4122A A_51_P301007 AB120716.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P301007 AK005610.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P301007 BC049742.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P301007 NM_029299.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P106595 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P111427 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07479546 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07479541 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07479542 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07479548 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07479543 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26375918 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26375885 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26375880 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26375875 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26375873 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26375916 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26375854 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26375877 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26375905 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26375846 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26375869 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26375915 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26375856 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26375849 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26375867 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26375858 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26375921 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26375851 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26375840 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26375907 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26375902 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26375866 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26375909 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26375845 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26375862 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26375904 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26375828 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26375882 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26375861 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26375826 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26375847 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26375836 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26375888 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26375910 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26375917 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26375913 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26375860 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26375837 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26375844 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26375865 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26375912 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26375883 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26375838 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07833599 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26375845 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07833598 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26375861 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08191664 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26375866 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26375913 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07743111 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26375885 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07743119 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26375847 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07739910 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26375877 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26375855 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26375860 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07990813 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07646930 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08138844 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26375837 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07891779 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26375865 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08191673 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26375918 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26375921 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26375838 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26375862 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26375844 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07698439 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26375875 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07646925 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26375834 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26375854 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26375873 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08191666 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26375858 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26375880 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26375869 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26375840 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26375888 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26375891 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26375836 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26375841 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26375912 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26375870 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26375851 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07758155 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26375915 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26375890 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26375848 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26375889 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10584020 - -
          Affymetrix Mouse Gene 1.0 ST Array 10584022 - -
          Affymetrix Mouse Gene 1.0 ST Array 10584016 - -
          Affymetrix Mouse Gene 1.0 ST Array 10584023 - -
          Affymetrix Mouse Gene 1.0 ST Array 10584021 - -
          Affymetrix Mouse Gene 1.0 ST Array 10584019 - -
          Affymetrix Mouse Gene 1.0 ST Array 10584017 - -
          Affymetrix HT MG-430 PM Array Plate 1417863_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10584015 - -
          Mouse Genome 430 2.0 Array 1417863_PM_at - -
          Mouse Genome 430 2.0 Array 1417863_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 75469_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 75469_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0900001985.mm.1 NM_029299.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0900001985.mm.1 TC0900000261.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900001986.mm.1 NM_029299.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0900001986.mm.1 TC0900000261.mm.1 MTA TC
          Mouse Expression Set 430 1417863_PM_at - -
          Mouse Expression Set 430 1417863_at - -
          Murine Genome U74v2 Set 165474_PM_f_at - -
          Murine Genome U74v2 Set 165474_f_at - -
          Mouse Genome 430A 2.0 Array 1417863_at - -
          Mouse Genome 430A 2.0 Array 1417863_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1417863_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1417863_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 424359 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 3140162 - -
          Illumina mouse-6 v1.0 expression beadchip 3140162 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003140162 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2636548 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2636548 - -
          mouse-6 v1.1 (Illumina) scl37179.8.1_83-S - -
          Illumina Sentrix mouse-6.v1 bead array 3140162 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl37179.8.1_83-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3140162 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2636548 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2636548 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl37179.8.1_83-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline