Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00504193_m1
          See other ABTB1 GE Assays ›
          Gene Symbol:
          Abtb1
          Gene Name:
          ankyrin repeat and BTB (POZ) domain containing 1
          Gene Aliases:
          AI847549, BC003234, BPOZ, EF1ABP
          Chromosome Location:
          Chr.6: 88835914 - 88842386 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00504193_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Abtb1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_030251.3 NP_084527.2 1 - 2 165 84
          GenBank mRNA AB053477.1 - 1 - 2 96 84
          AK088437.1 - 1 - 2 130 84
          AK155432.1 - 1 - 2 118 84
          AK155505.1 - 1 - 2 118 84
          AK155644.1 - 1 - 2 118 84
          BB850379.1 - 1 - 2 168 84
          BC003234.1 - 1 - 2 109 3483893 84
          BY201769.1 - 1 - 2 123 84

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600002688.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Abtb1

          Gene Ontology Categories:

          Function(s) Process(es)

          translation
          translational elongation
          biological_process
          molecular_function
          translation elongation factor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24624506 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24624508 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24624507 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24624509 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24624502 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24624504 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24624511 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24624505 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24624503 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1979217 AB053477.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1979217 NM_030251.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 749856 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 902642 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 939404 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 99120 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P121905 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1979217 AB053477.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1979217 NM_030251.3 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24624491 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24624494 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24624507 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24624490 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24624487 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24624511 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24624500 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24624502 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24624495 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24624505 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24624489 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24624510 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24624503 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24624496 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24624484 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24624501 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24624483 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24624498 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24624509 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24624493 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01379913 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01379905 - -
          Whole Mouse Genome G4122A A_51_P267671 AK220208.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P267671 AK155505.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P267671 AK155432.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P267671 AK155644.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P267671 BC003234.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P101151 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05357098 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05357061 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05357053 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05357088 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24624506 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24624509 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24624502 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24624505 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24624507 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24624504 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24624503 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24624511 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24624508 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24624435 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24624461 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24624495 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24624450 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24624440 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24624482 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24624463 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24624489 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24624447 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24624448 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24624507 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24624510 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24624496 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24624484 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24624443 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24624500 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24624511 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24624502 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24624480 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24624491 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24624487 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24624429 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24624473 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24624477 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24624478 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24624441 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24624475 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24624452 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24624465 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24624509 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24624459 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24624470 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24624425 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24624438 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24624445 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24624430 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24624454 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24624498 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24624456 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24624472 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24624432 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24624503 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24624433 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24624467 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24624469 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24624426 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24624464 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24624493 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24624494 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24624505 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05470018 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24624474 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24624494 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24624456 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05470028 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05469997 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05564131 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24624491 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24624450 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24624480 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05564125 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05564129 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24624498 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24624507 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05564118 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05470027 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24624505 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05470030 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05470010 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05357098 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05357088 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05469998 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24624496 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24624490 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05564117 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24624502 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24624469 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24624463 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05470012 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05564106 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05470031 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24624489 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24624500 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24624509 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24624475 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05564126 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24624443 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05564112 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05470020 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24624448 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24624464 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24624438 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05564130 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10546138 - -
          Affymetrix Mouse Gene 1.0 ST Array 10546139 - -
          Affymetrix Mouse Gene 1.0 ST Array 10546146 - -
          Affymetrix Mouse Gene 1.0 ST Array 10546142 - -
          Affymetrix Mouse Gene 1.0 ST Array 10546141 - -
          Affymetrix Mouse Gene 1.0 ST Array 10546144 - -
          Affymetrix Mouse Gene 1.0 ST Array 10546151 - -
          Affymetrix Mouse Gene 1.0 ST Array 10546145 - -
          Affymetrix Mouse Gene 1.0 ST Array 10546140 - -
          Affymetrix Mouse Gene 1.0 ST Array 10546148 - -
          Affymetrix Mouse Gene 1.0 ST Array 10546147 - -
          Affymetrix Mouse Gene 1.0 ST Array 10546150 - -
          Affymetrix Mouse Gene 1.0 ST Array 10546149 - -
          Affymetrix Mouse Gene 1.0 ST Array 10546143 - -
          Affymetrix HT MG-430 PM Array Plate 1417000_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10546137 - -
          Mouse Genome 430 2.0 Array 1417000_at - -
          Mouse Genome 430 2.0 Array 1417000_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 80283_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 80283_at - -
          Affymetrix Mouse Clariom S Array PSR0600020905.mm.1 TC0600002688.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600020905.mm.1 NM_030251.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600020905.mm.1 NM_030251.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0600020906.mm.1 TC0600002688.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600020906.mm.1 NM_030251.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600020906.mm.1 NM_030251.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0600020907.mm.1 TC0600002688.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600020907.mm.1 NM_030251.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600020907.mm.1 NM_030251.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0600020909.mm.1 TC0600002688.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600020909.mm.1 NM_030251.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600020909.mm.1 NM_030251.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0600020908.mm.1 TC0600002688.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600020908.mm.1 NM_030251.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600020908.mm.1 NM_030251.3 Refseq
          Mouse Expression Set 430 1417000_PM_at - -
          Mouse Expression Set 430 1417000_at - -
          Mouse Genome 430A 2.0 Array 1417000_PM_at - -
          Mouse Genome 430A 2.0 Array 1417000_at - -
          Affymetrix HT Mouse Genome 430A Array 1417000_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1417000_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 559176 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 4780086 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004780086 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2699477 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2699477 - -
          mouse-6 v1.1 (Illumina) scl28709.9_141-S - -
          Illumina Sentrix mouse-6.v1 bead array 4780086 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl28709.9_141-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4780086 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2699477 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2699477 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl28709.9_141-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline