Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00504282_m1
          See other EIF4H GE Assays ›
          Gene Symbol:
          Eif4h
          Gene Name:
          eukaryotic translation initiation factor 4H
          Gene Aliases:
          AU018978, D5Ertd355e, E430026L18Rik, Ef4h, Wbscr1, Wscr1, mKIAA0038
          Chromosome Location:
          Chr.5: 134619876 - 134639494 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00504282_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Eif4h
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_033561.1 NP_291039.1 1 - 2 88 90
          GenBank mRNA AK045004.1 - 1 - 2 90 90
          AK088818.1 - 1 - 2 100 90
          AK122194.1 - 1 - 2 88 90
          AK132034.1 - 1 - 2 108 90
          AK135390.1 - 1 - 2 89 90
          AK136999.1 - 1 - 2 75 90
          AK149841.1 - 1 - 2 74 90
          AK150088.1 - 1 - 2 97 90
          AK166567.1 - 1 - 2 89 90
          AK168872.1 - 1 - 2 107 90
          AK169312.1 - 1 - 2 71 90
          AK169353.1 - 1 - 2 89 90
          AK169407.1 - 1 - 2 107 90
          BC014796.1 - 1 - 2 102 3962104 90

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500003377.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Eif4h

          Panther Classification:

          Molecular Function -

          translation initiation factor

          Gene Ontology Categories:

          Function(s) Process(es)

          translation
          translational initiation
          sexual reproduction
          developmental growth
          nucleotide binding
          nucleic acid binding
          RNA binding
          translation initiation factor activity
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32982443 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32982439 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32982441 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32982436 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32982435 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32982444 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1974582 BC014796.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1974582 BC023227.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1974582 BC023337.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1974582 AK122194.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1974582 AK169407.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 101642 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 455257 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 908521 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 58218 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 851937 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 701421 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 908752 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 926238 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 861133 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 255324 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 585068 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 178046 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 151532 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1974582 BC014796.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1974582 BC023227.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1974582 BC023337.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1974582 AK122194.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1974582 AK169407.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P228398 BC014796.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P228398 BC023227.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P228398 BC023337.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P228398 AK122194.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P228398 AK169407.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P125776 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P140943 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32982436 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32982435 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32982439 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32982443 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32982441 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32982444 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P32982358 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P32982421 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P32982343 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P32982416 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P32982355 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P32982354 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P32982353 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P32982339 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P32982419 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P32982407 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P32982430 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P32982341 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P32982411 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P32982347 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P32982426 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P32982423 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P32982443 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P32982348 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P32982424 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P32982427 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P32982345 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P32982425 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P32982335 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P32982417 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P32982342 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P32982412 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P32982429 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P32982349 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P32982332 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P32982337 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P32982414 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P32982338 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P32982446 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P32982331 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P32982359 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P32982352 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P32982357 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P32982356 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P32982438 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P32982433 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P32982422 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P32982406 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P32982435 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P32982413 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P32982351 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P32982449 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P32982450 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P32982409 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P32982445 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10534319 - -
          Affymetrix Mouse Gene 1.0 ST Array 10534320 - -
          Affymetrix Mouse Gene 1.0 ST Array 10534317 - -
          Affymetrix Mouse Gene 1.0 ST Array 10534321 - -
          Affymetrix Mouse Gene 1.0 ST Array 10534318 - -
          Affymetrix Mouse Gene 1.0 ST Array 10534322 - -
          Affymetrix Mouse Gene 1.0 ST Array 10534323 - -
          Affymetrix HT MG-430 PM Array Plate 1445746_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1451521_PM_x_at - -
          Affymetrix HT MG-430 PM Array Plate 1416941_PM_s_at - -
          Affymetrix HT MG-430 PM Array Plate 1447070_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1442942_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1438554_PM_x_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10534316 - -
          Mouse Genome 430 2.0 Array 1442942_at - -
          Mouse Genome 430 2.0 Array 1436637_at - -
          Mouse Genome 430 2.0 Array 1416941_s_at - -
          Mouse Genome 430 2.0 Array 1445746_PM_at - -
          Mouse Genome 430 2.0 Array 1416941_PM_s_at - -
          Mouse Genome 430 2.0 Array 1451521_PM_x_at - -
          Mouse Genome 430 2.0 Array 1442942_PM_at - -
          Mouse Genome 430 2.0 Array 1445746_at - -
          Mouse Genome 430 2.0 Array 1438554_x_at - -
          Mouse Genome 430 2.0 Array 1438554_PM_x_at - -
          Mouse Genome 430 2.0 Array 1436637_PM_at - -
          Mouse Genome 430 2.0 Array 1451521_x_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 22384_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 22384_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0500030454.mm.1 TC0500003377.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500030454.mm.1 NM_033561.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500030452.mm.1 TC0500003377.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500030452.mm.1 NM_033561.2 Refseq
          Mouse Expression Set 430 1451521_PM_x_at - -
          Mouse Expression Set 430 1438554_PM_x_at - -
          Mouse Expression Set 430 1451521_x_at - -
          Mouse Expression Set 430 1416941_PM_s_at - -
          Mouse Expression Set 430 1416941_s_at - -
          Mouse Expression Set 430 1438554_x_at - -
          Mouse Expression Set 430 1445746_PM_at - -
          Mouse Expression Set 430 1445746_at - -
          Mouse Expression Set 430 1442942_PM_at - -
          Mouse Expression Set 430 1442942_at - -
          Murine Genome U74v2 Set 97278_at - -
          Murine Genome U74v2 Set 97278_PM_at - -
          Mouse Genome 430A 2.0 Array 1438554_x_at - -
          Mouse Genome 430A 2.0 Array 1416941_s_at - -
          Mouse Genome 430A 2.0 Array 1438554_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1451521_x_at - -
          Mouse Genome 430A 2.0 Array 1416941_PM_s_at - -
          Mouse Genome 430A 2.0 Array 1451521_PM_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1438554_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1438554_PM_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1451521_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1416941_PM_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1451521_PM_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1416941_s_at - -
          Affymetrix HT Mouse Genome 430B Array 1442942_at - -
          Affymetrix HT Mouse Genome 430B Array 1442942_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1447070_at - -
          Affymetrix HT Mouse Genome 430B Array 1445746_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1447070_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1445746_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 922881 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 610324 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000610324 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1243395 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1243395 - -
          mouse-6 v1.1 (Illumina) scl25941.7_8-S - -
          Illumina Sentrix mouse-6.v1 bead array 610324 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl25941.7_8-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 610324 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1243395 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1243395 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl25941.7_8-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-t4kr2:80/100.66.77.141:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0