Search
Search
| Gene Symbol: Bcl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_033601.3 | NP_291079.2 | 1 - 2 | 325 | 83 | |
| GenBank mRNA | AF067774.1 | - | 1 - 2 | 286 | 83 | |
| AK155016.1 | - | 1 - 2 | 290 | 83 | ||
| BY749796.1 | - | 1 - 2 | 326 | 83 | ||
| M90397.1 | - | 1 - 2 | 243 | 83 | ||
Set Membership: |
|