Search
Search
| Gene Symbol: Ric8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053194.4 | NP_444424.1 | 5 - 6 | 1040 | 62 | |
| GenBank mRNA | AF288813.1 | - | 5 - 6 | 1039 | 62 | |
| AK031466.1 | - | 5 - 6 | 987 | 62 | ||
| AK080192.1 | - | 5 - 6 | 983 | 62 | ||
| AK155282.1 | - | 5 - 6 | 1015 | 62 | ||
| AK165536.1 | - | 5 - 6 | 1017 | 62 | ||
| AK167746.1 | - | 5 - 6 | 1043 | 62 | ||
| AK169367.1 | - | 5 - 6 | 1004 | 62 | ||
| BC022917.1 | - | 5 - 6 | 1017 | 4460521 | 62 | |
Set Membership: |
|